Skip to Content
Merck
All Photos(1)

Key Documents

343290

Sigma-Aldrich

Fluoxetine hydrochloride

≥98% (HPLC), solid, selective serotonin re-uptake inhibitor, Calbiochem

Synonym(s):

Fluoxetine, Hydrochloride, dl-N-Methyl-3-( p-trifluoromethylphenoxy)-3-phenylpropylamine, HCl, (±)-N-Methyl-3-phenyl-3-[(α,α,α-trifluoro- p-tolyl)oxy]propylamine, HCl, LY-110140, Prozac

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$459.00

$459.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$459.00

About This Item

Empirical Formula (Hill Notation):
C17H18F3NO
CAS Number:
Molecular Weight:
309.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$459.00


Check Cart for Availability

Request a Bulk Order

Product Name

Fluoxetine, Hydrochloride, A cell-permeable phenylpropylamine derived anti-depressant that acts as a selective serotonin re-uptake inhibitor (SSRI).

Quality Level

description

Merck USA index - 14, 4185

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

methanol: 100 mg/mL
water: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1ccc(cc1)OC(CCNC)c2ccccc2.[Cl-].[H+]

InChI

1S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21H,11-12H2,1H3;1H

InChI key

GIYXAJPCNFJEHY-UHFFFAOYSA-N

General description

A cell-permeable phenylpropylamine derived anti-depressant that acts as a selective serotonin re-uptake inhibitor (SSRI). Reported to modulate the proliferation of T cells by increasing the Ca2+ influx thereby influencing the activities of protein kinase A (PKA) and protein kinase C (PKC). Inhibits leak K+ currents in a reversible and voltage-independent manner (IC50 = 780 nM). Also shown to regulate the phosphorylation of DARPP-32 (dopamine-and cAMP-regulated phosphoprotein of Mr 32,000) and AMPA (α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid) receptors. Triggers oxidative stress-induced apoptotic cell death and initiates signaling pathways in neuronal cells leading to an activation of ERK and NF-κB.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Selective serotonin re-uptake inhibitor
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bartholoma, P., et al. 2002. Biochem. Pharmacol.63, 1507.
Bian, J.T., et al. 2002. Eur. J. Pharmacol.453, 159.
Zhang, A., et al. 2002. J. Med. Chem.45, 1930.
Svenningsson, P., et al. 2002. Proc. Natl. Acad. Sci. USA99, 3182.
Edgar, V.A., et al. 1999. Eur. J. Pharmacol.372, 65.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Is 343290 a racemic mixture?

    1 answer
    1. Yes, this is a racemic mixture. This can be inferred from the synonyms, which include dl and +/-. For example, Fluoxetine, Hydrochloride, dl-N-Methyl-3-(p-trifluoromethylphenoxy)-3-phenylpropylamine, HCl, (±)-N-Methyl-3-phenyl-3-[(α,α,α-trifluoro-p-tolyl)oxy]propylamine, HCl, LY-110140, Prozac.

      Helpful?

  2. Is this a racemic mixture?

    1 answer
    1. As per the PDP, this product is a racemic mixture.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service