Skip to Content
Merck
All Photos(1)

Documents

343209

Sigma-Aldrich

Fluorescein Diacetate

≥99% (HPLC), solid, esterase substrate, Calbiochem®

Synonym(s):

Fluorescein Diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16O7
CAS Number:
Molecular Weight:
416.38
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.77

product name

Fluorescein Diacetate, Stains living cells green. Substrate for esterases. Suitable for use in cell viability assays. Removal of the acetate groups by cellular esterases results in fluorescence in cells with intact membranes.

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

colorless to yellow

solubility

ethanol: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C24H16O7/c1-13(25)28-15-7-9-19-21(11-15)30-22-12-16(29-14(2)26)8-10-20(22)24(19)18-6-4-3-5-17(18)23(27)31-24/h3-12H,1-2H3

InChI key

CHADEQDQBURGHL-UHFFFAOYSA-N

General description

Stains living cells green. Substrate for esterases. Suitable for use in cell viability assays. Removal of the acetate groups by cellular esterases results in fluorescence in cells with intact membranes.
Substrate for lipases and esterases. Abs. Max: 498 nm.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution. Reconstitute just prior to use.

Other Notes

Clements, K.M., et al. 2001. Osteoarthritis Cartilage9, 499.
Bedner, E., et al. 1998. Cytometry33, 1.
Guilbault, G.G., and Kramer, D.N. 1964. Anal. Chem. 36, 409.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service