Skip to Content
Merck
All Photos(1)

Key Documents

341608

Sigma-Aldrich

FGF Receptor Tyrosine Kinase Inhibitor

The FGF Receptor Tyrosine Kinase Inhibitor, also referenced under CAS 192705-79-6, controls the biological activity of FGF Receptor Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

FGF Receptor Tyrosine Kinase Inhibitor, 1-(2-Amino-6-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-7-yl)-3-tert-butyl urea, PD 166866, PD 166866, 1-(2-Amino-6-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-7-yl)-3-tert-butyl urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N6O3
CAS Number:
Molecular Weight:
396.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Product 341608 is not currently sold in your country. Contact Technical Service

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(C(C)(C)C)C(=O)Nc1nc2nc(ncc2cc1c3cc(cc(c3)OC)OC)N

InChI

1S/C20H24N6O3/c1-20(2,3)26-19(27)25-17-15(8-12-10-22-18(21)24-16(12)23-17)11-6-13(28-4)9-14(7-11)29-5/h6-10H,1-5H3,(H4,21,22,23,24,25,26,27)

InChI key

NHJSWORVNIOXIT-UHFFFAOYSA-N

General description

A cell-permeable pyridopyrimidinyl urea compound that acts a potent, ATP-competitive inhibitor of FGFR (IC50 = 60 nM using FGFR1) with excellent selectivity over PDGFRβ, EGFR, insulin receptor, and c-Src (IC50 >50 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Panek, R.L., et al. 1998. J. Pharm. Exp. Ther.286, 569.
Hamby, J.M., et al. 1997. J. Med. Chem.40, 2296.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander S Brown et al.
Development (Cambridge, England), 138(18), 3967-3976 (2011-08-13)
In mouse embryos lacking sonic hedgehog (Shh), dorsoventral polarity within the otic vesicle is disrupted. Consequently, ventral otic derivatives, including the cochlear duct and saccule, fail to form, and dorsal otic derivatives, including the semicircular canals, endolymphatic duct and utricle

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service