Skip to Content
Merck
All Photos(1)

Key Documents

341305

Sigma-Aldrich

Fasentin

Synonym(s):

Fasentin, 6094911, N-(4-Chloro-3-(trifluoromethyl)phenyl)-3-oxobutanamide, Glucose Transporter Inhibitor, Glucose Transporter Inhibitor, 6094911, N-(4-Chloro-3-(trifluoromethyl)phenyl)-3-oxobutanamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$218.00

$218.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$218.00

About This Item

Empirical Formula (Hill Notation):
C11H9ClF3NO2
CAS Number:
Molecular Weight:
279.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

$218.00


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(F)(F)c1c(ccc(c1)NC(=O)CC(=O)C)Cl

InChI

1S/C11H9ClF3NO2/c1-6(17)4-10(18)16-7-2-3-9(12)8(5-7)11(13,14)15/h2-3,5H,4H2,1H3,(H,16,18)

InChI key

GNYIJZMBLZXJEJ-UHFFFAOYSA-N

General description

A cell-permeable oxobutananilide compound that is predicted to interact with Glut (Glucose Transporter) at an intracellular site based on a virtual docking study and shown to inhibit Glut-mediated cellular glucose uptake (50% and 80% inhibition of 2-deoxy-D-glucose uptake in Glut1- and Glut4-overexpressing L6 myoblasts, respectively, at 300 µM). Fas-resistant cell lines that are made Fas-responsive upon medium glucose deprivation are also shown to be sensitized by Fasentin to Fas-mediated cell death (ED50 = 35 µM in PPC-1 cultures), while cells that remain Fas-resistant upon glucose deprivation, e.g. DU145, are shown not to be sensitized by Fasentin to Fas-mediated death.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -2°C.

Other Notes

Wood, T.E., et al. 2008. Mol. Cancer Ther.7, 3546
Schimmer, A.D., et al. 2006. Cancer Res.66, 2367.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service