Skip to Content
Merck
All Photos(1)

Key Documents

328011

Sigma-Aldrich

ERK Inhibitor VIII

VX-11e

Synonym(s):

ERK Inhibitor VIII

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$398.00

$398.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$398.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

$398.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

SMILES string

Fc1cc(c(cc1)Nc2nc(c(cn2)C)c3c[nH]c(c3)C(=O)N[C@H](CO)c4cc(ccc4)Cl)Cl

InChI key

WUTVMXLIGHTZJC-OAQYLSRUSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service