Skip to Content
Merck
All Photos(1)

Key Documents

328000

Sigma-Aldrich

ERK Activation Inhibitor Peptide I, Cell-Permeable

The ERK Activation Inhibitor Peptide I, Cell-Permeable controls the biological activity of ERK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

ERK Activation Inhibitor Peptide I, Cell-Permeable, Ste-MEK1₁₃, Ste-MPKKKPTPIQLNP-NH₂, Erk Inhibitor IV

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$338.00

$338.00


Estimated to ship on30 May 2025



Select a Size

Change View
1 MG
$338.00

About This Item

Empirical Formula (Hill Notation):
C86H153N19O17S
Molecular Weight:
1757.32
UNSPSC Code:
12352200
NACRES:
NA.77

$338.00


Estimated to ship on30 May 2025


Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

potency

2.5 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable and reversible stearated 13-amino acid peptide corresponding to the N-terminus of MEK1 (MAPKK) that acts as a specific inhibitor of ERK activation and blocks the transcriptional activity of Elk1. Selectively binds to ERK2 and prevents its interaction with MEK (IC50 = 2.5 µM). Potently blocks ERK activation in PMA-stimulated NIH 3T3 cells and NGF-treated PC12 cells (IC50 = 13 µM). Does not affect the activation of JNKs or p38.
A cell-permeable, reversible, and stearated, 13-amino acid peptide corresponding to the N-terminus of MAPK/ERK kinase (MEK). Acts as a specific inhibitor of ERK activation and blocks the transcriptional activity of ELK1. Selectively binds to ERK2 and prevents its interaction with MEK (IC50 = 2.5 µM). Shown to block ERK activity in PMA-stimulated NIH3T3 cells and NGF-treated PC12 cells (IC50 = 13 µM). Does not affect the activation of either JNKs or p38 MAP kinase.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MEK
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Ste-Met-Pro-Lys-Lys-Lys-Pro-Thr-Pro-Ile-Gln-Leu-Asn-Pro-NH₂ (Ste = stearated)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kelemen, B.R., et al. 2002. J. Biol. Chem.277, 8741.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Fang et al.
International journal of molecular sciences, 23(22) (2022-11-27)
Chronic obstructive pulmonary disease (COPD) is characterized by irreversible deterioration of the airway wall. Cigarette smoking is the major trigger, and in vitro studies showed that cigarette smoke extract (CSE) induced mitophagy in airway epithelial cells via oxidative stress, but
Lena Wiedmann et al.
Nature communications, 14(1), 2353-2353 (2023-04-25)
Pancreatic ductal adenocarcinoma (PDAC) frequently metastasizes into the peritoneum, which contributes to poor prognosis. Metastatic spreading is promoted by cancer cell plasticity, yet its regulation by the microenvironment is incompletely understood. Here, we show that the presence of hyaluronan and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service