Skip to Content
Merck
All Photos(1)

Key Documents

324888

Sigma-Aldrich

Epigenetic Multiple Ligand

A cell-permeable bis-arylidene compound that acts as an epigenetic multiple ligand (epi-ML) and inhibits substrates processing by several chromatin-associated enzymes.

Synonym(s):

Epigenetic Multiple Ligand, 3,5-bis(3,5-Dibromo-4-hydroxybenzylidene)-tetrahydro-2H-pyran-4-one, PRMT Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12Br4O4
CAS Number:
Molecular Weight:
623.91
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light orange

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable bis-arylidene compound that acts as an epigenetic multiple ligand (epi-ML) and inhibits substrates processing by several chromatin-associated enzymes, including H4/PRMT1 (91% inhibition at 50 µM), H4/RmtA (50% inhibition at 29 µM), H3/p300/CBP (61% inhibition at 50 µM), PABP1/CARM1 (>90% inhibition at 100 µM), H3/SET7 (>90% inhibition at 100 µM), SIRT1/2 (>50% inhibition at 25 µM), although the process of Np13p by PRMT1 is only weakly affected. Shown to induce either apoptosis (28% after 30 h incubation with 25 µM inhibitor) or granulocytic differentiation (100% of non-apoptotic cells after 30 hr incubation with 5 µM inhibitor) among U937 cell population.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Mai, A., et al. 2008. J. Med. Chem.51, 2279.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service