Skip to Content
Merck
All Photos(1)

Key Documents

324831

Sigma-Aldrich

EGF human

≥95% (SDS-PAGE), recombinant, expressed in E. coli, solid, suitable for cell culture

Synonym(s):

EGF, Human, Recombinant, E. coli, Epidermal Growth Factor, Human, Recombinant, E. coli

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available. Contact Technical Service

Product Name

EGF, Human, Recombinant, E. coli,

recombinant

expressed in E. coli

Quality Level

description

Merck USA index - 14, 3527

Assay

≥95% (SDS-PAGE)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg Endotoxin (EU/μg EGF)

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, human epidermal growth factor (54 amino acids) expressed in E. coli. Identical to native human EGF except for an additional N-terminal methionine. Human EGF is 70% homologous to mouse EGF and has been shown to have similar biological activity. Mitogenic for a wide range of ectoderm- and endoderm-derived cells from tissue such as breast, cornea, epidermis, dermis, liver, pancreas, nerve, amnion, and adrenal medulla. Binding of EGF to its 170 kDa transmembrane receptor results in dimerization of the receptor, activation of the intrinsic receptor tyrosine kinase, and initiation of the complex signal transduction cascade that leads to cell proliferation and/or differentiation. Acts as a survival factor in preventing apoptosis.

Biochem/physiol Actions

ED₅₀ = 100-400 pg/ml as measured by its ability to stimulate the ³H-thymidine incorporation in an EGF-responsive mouse fibroblast cell line, Balb/3T3.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C or -70°C) for long term storage or refrigerate (4°C) for short-term storage. Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C or -70°C.
Reconstitute to a concentration of ≥10 µg/ml in 10 mM acetic acid containing ≥0.1% HSA or BSA.

Other Notes

Merlo, G.R., et al. 1995. J. Cell Biol. 128, 1185.
Yoshida, T., et al. 1993. Brain Res. Dev.76, 147.
Rubin, J.S., et al. 1991. Proc. Natl. Acad. Sci. USA88, 415.
Carpenter, G., and Wahl, M.I. 1990. In The Epidermal Growth Factor Family in Peptide Growth Factors and Their Receptors (Sporn, M.B., and Roberts, A.B., eds.) Springer-Verlag, New York, p 69.
Downward, J., et al. 1984. Nature311, 483.
Gray, A., et al. 1983. Nature303, 722.
Scott, J., et al. 1983. Science221, 236.
Cohen, S. 1960. Proc. Natl. Acad. Sci. USA46, 302.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Okerberg et al.
Current protocols in chemical biology, 5(3), 213-226 (2014-01-07)
The protocols in this unit describe efficient and cost-effective approaches to determine the interaction of small-molecule inhibitors with native kinases, and also analyze the interactions between kinases and their binding partners in a cellular setting. The combined attributes of activity-based
Katarzyna A Ludwik et al.
Cell reports, 32(3), 107931-107931 (2020-07-23)
In response to estrogens, estrogen receptor alpha (ERα), a critical regulator of homeostasis, is degraded through the 26S proteasome. However, despite the continued presence of estrogen before menopause, ERα protein levels are maintained. We discovered that ERK1/2-RSK2 activity oscillates during
Marta Grońska-Pęski et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 41(13), 2899-2910 (2021-02-28)
The addition of new neurons to existing neural circuits in the adult brain remains of great interest to neurobiology because of its therapeutic implications. The premier model for studying this process has been the hippocampal dentate gyrus in mice, where
Lindsey Seldin et al.
Developmental cell, 55(5), 558-573 (2020-10-16)
DNA crosslinking agents are commonly used in cancer chemotherapy; however, responses of normal tissues to these agents have not been widely investigated. We reveal in mouse interfollicular epidermal, mammary and hair follicle epithelia that genotoxicity does not promote apoptosis but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service