Skip to Content
Merck
All Photos(1)

Key Documents

324483

Sigma-Aldrich

Ebselen

Synonym(s):

Ebselen, 2-Phenyl-1,2-benzisoselenazol-3(2H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$166.00

$166.00


Estimated to ship on05 June 2025FromTruganina


Request a Bulk Order

Select a Size

Change View
5 MG
$166.00

About This Item

Empirical Formula (Hill Notation):
C13H9NOSe
CAS Number:
Molecular Weight:
274.18
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

$166.00


Estimated to ship on05 June 2025FromTruganina


Request a Bulk Order

Quality Level

SMILES string

O=C1N([Se]c2ccccc12)c3ccccc3

InChI

1S/C13H9NOSe/c15-13-11-8-4-5-9-12(11)16-14(13)10-6-2-1-3-7-10/h1-9H

InChI key

DYEFUKCXAQOFHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service