Skip to Content
Merck
All Photos(1)

Key Documents

322338

Sigma-Aldrich

Dvl-PDZ Domain Inhibitor II

The Dvl-PDZ Domain Inhibitor II, also referenced under CAS 294891-81-9, controls the biological activity of Dvl-PDZ. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Dvl-PDZ Domain Inhibitor II, Wnt Pathway Inhibitor IV, 2-((3-(2-Phenylacetyl)amino)benzoyl)amino)benzoic acid, 3289-8625

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$202.00

$202.00


Estimated to ship on29 April 2025



Select a Size

Change View
10 MG
$202.00

About This Item

Empirical Formula (Hill Notation):
C22H18N2O4
CAS Number:
Molecular Weight:
374.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

$202.00


Estimated to ship on29 April 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(c2cc(ccc2)C(=O)Nc3c(cccc3)C(=O)O)C(=O)Cc1ccccc1

InChI

1S/C22H18N2O4/c25-20(13-15-7-2-1-3-8-15)23-17-10-6-9-16(14-17)21(26)24-19-12-5-4-11-18(19)22(27)28/h1-12,14H,13H2,(H,23,25)(H,24,26)(H,27,28)

InChI key

OBGIRQUECFHUEY-UHFFFAOYSA-N

General description

A cell-permeable amidobenzanilide compound that disrupts Fz-Dvl (frizzled-dishevelled) interaction by targeting the PDZ domain (Kd = 10.6 µM) of Dvl, blocking Wnt3a-induced (10 ng/ml) transcription activity (~50% inhibition at 3 µM in 293 cell SuperTopflash reporter assays) and suppressing the Wnt pathway-dependent growth of prostate cancer PC-3 cells (by 16% in 72 h at ≥50 µM; IC50 = 12.5 µM). Shown to effectively prevent Wnt3A mRNA injection-induced xenopus embryo deformation (effective conc. = 25 µM) and suppress Wnt pathway-mediated apoptosis of the hyaloid vescular endothelial cells in the mouse eyes via vitreous injection (144 fmol/120 nl/eye) in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Grandy, D., et al. 2009. J. Biol. Chem.284, 16256.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yisong Xu et al.
Artificial cells, nanomedicine, and biotechnology, 46(sup2), 192-200 (2018-03-28)
Acquired resistance to 5-fluorouracil (5-FU) frequently occurs in patients with hepatocellular carcinoma (HCC), the underlying molecular mechanisms of which are poorly understood. The aim of this study was to identify candidate genes and associated signalling pathways that may play a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service