Skip to Content
Merck
All Photos(1)

Documents

309866

Sigma-Aldrich

Diltiazem, Hydrochloride

Synthetic benzothiazepine that acts as an L-type Ca2+ channel antagonist.

Synonym(s):

Diltiazem, Hydrochloride, L-Type Calcium Channel Blocker II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26N2O4S · xHCl
CAS Number:
Molecular Weight:
414.52 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 50 mg/mL
chloroform: soluble
methanol: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C22H26N2O4S.ClH/c1-15(25)28-20-21(16-9-11-17(27-4)12-10-16)29-19-8-6-5-7-18(19)24(22(20)26)14-13-23(2)3;/h5-12,20-21H,13-14H2,1-4H3;1H/t20-,21+;/m1./s1

InChI key

HDRXZJPWHTXQRI-BHDTVMLSSA-N

General description

Synthetic (+)-cis isomer that acts as an L-type Ca2+ channel antagonist. Similar in function to nifedipine and verapamil. Has a negative inotropic effect on the heart. Strongly suppresses conductance at the A-V node and automatically at the S-A node. Has also been shown to regulate Ca2+ release from intracellular stores in neutrophils.
Synthetic benzothiazepine that acts as an L-type Ca2+ channel antagonist. Similar in function to Nifedipine (Cat. No. 481981) and Verapamil (Cat. No. 676777). Has negative inotropic effects on the heart. Strongly suppresses conductance at the A-V node and automaticity at the S-A node. Has also been shown to regulate Ca2+ release from intracellular stores in neutrophils.

Biochem/physiol Actions

Cell permeable: no
Primary Target
L-type Ca2+ channel
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 2 weeks at 4°C.

Other Notes

van Zwieten, P.A., and Pfaffendorf, M. 1993. J. Hypertens. Suppl. 11, S3.
Rosales, C., and Brown, E.J. 1992. J. Biol. Chem. 267, 1443.
Julian, D.G. 1987. Am. J. Cardiol. 59, 37J.
Campbell, K.P., et al. 1986. Proc. Natl. Acad. Sci. USA83, 2792.
Gallizzi, J.P., et al. 1986. J. Biol. Chem.261, 1393.
Garcia, M.L., et al. 1986. J. Biol. Chem.261, 8146.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service