Skip to Content
Merck
All Photos(1)

Key Documents

300267

Sigma-Aldrich

DPPH, Free Radical

≥90% (elemental analysis), solid, hydrogen radical scavenger, Calbiochem®

Synonym(s):

DPPH, Free Radical

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$159.00

$159.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
50 MG
$159.00

About This Item

Empirical Formula (Hill Notation):
C18H12N5O6
CAS Number:
Molecular Weight:
394.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

$159.00


In StockDetails


Request a Bulk Order

Product Name

DPPH, Free Radical, DPPH, Free Radical, CAS 1898-66-4, is a cell-permeable, stable free radical that acts as a hydrogen radical scavenger.

Quality Level

Assay

≥90% (elemental analysis)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

black

solubility

DMF: 10 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[N+](=O)([O-])c1c(c(cc(c1)[N+](=O)[O-])[N+](=O)[O-])NN(c3ccccc3)c2ccccc2

InChI

1S/C18H13N5O6/c24-21(25)15-11-16(22(26)27)18(17(12-15)23(28)29)19-20(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-12,19H

InChI key

WCBPJVKVIMMEQC-UHFFFAOYSA-N

General description

A cell-permeable, stable free radical that acts as a hydrogen radical scavenger. Useful in inducing free-radical injury to tissues and as a screening tool for detecting free radical scavenging activity of antioxidants. Absorption max: ~517 nm.
A cell-permeable, stable, free radical that acts as a hydrogen radical (H) scavenger. Useful to induce free-radical injury to tissues and as a screening tool for detecting antioxidant activity. Reduction in the levels of DPPH can be used as a measure of antioxidant properties of compounds. Note: Absorption max: ~ 517 nm.
DPPH, Free Radical, CAS 1898-66-4, is a cell-permeable, stable free radical that acts as a hydrogen radical scavenger.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Hydrogen radical scavenger
Product does not compete with ATP.

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Unstable in aqueous solutions.

Other Notes

Weber, V., et al. 2001. Arzneimittelforschung51, 877.
Karki, S.B., et al. 2000. J. Pharm. Sci.89, 1518.
Ancerewicz, J., et al. 1998. Free Radic. Biol. Med.25, 113.
Dinis, T.C., et al. 1994. Arch. Biochem. Biophys.315, 161.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Self-react. C - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service