Skip to Content
Merck
All Photos(1)

Key Documents

251406

Sigma-Aldrich

D-Amino Acid Oxidase Inhibitor III

AS057278

Synonym(s):

3-Methylpyrazole-5-carboxylic acid, 5-Methylpyrazole-3-carboxylic acid, AS057278, MPC

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$202.00

$202.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$202.00

About This Item

Empirical Formula (Hill Notation):
C5H6N2O2
CAS Number:
Molecular Weight:
126.11
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.28

$202.00


Estimated to ship on20 May 2025


Request a Bulk Order

Quality Level

mp

236-240 °C (lit.)

SMILES string

Cc1cc([nH]n1)C(O)=O

InChI

1S/C5H6N2O2/c1-3-2-4(5(8)9)7-6-3/h2H,1H3,(H,6,7)(H,8,9)

InChI key

WSMQKESQZFQMFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service