Skip to Content
Merck
All Photos(1)

Key Documents

239799

Sigma-Aldrich

Ceruloplasmin, Human Plasma

Ceruloplasmin, Human Plasma, CAS 9031-37-2, from human plasma is involved in serum copper transport and plays a protective role against oxygen radicals generated by iron and ascorbate.

Synonym(s):

Ceruloplasmin, Human Plasma

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$712.00

$712.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$712.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

$712.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

human plasma

Quality Level

description

Merck USA index - 14, 2006

Assay

≥95% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

pI 

4.4

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Native ceruloplasmin from human plasma. Serum copper transport and iron-oxidizing protein. Plays an important role in antioxidant protection against organic and inorganic oxygen radicals generated by iron and ascorbate.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 100 mM KCl, 50 mM potassium phosphate buffer, 20 mM ε-aminocaproic acid, 5 mM EDTA, pH 6.8.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock Solutions are stable for up to 4 months at -20°C.

Other Notes

Gutteridge, J.M., and Quinlan, G.J. 1993. Biochim. Biophys. Acta1156, 144.
Krsek-Staples, J.A., and Webster, R.O. 1993. Free Radic. Biol. Med.14, 115.
Harris, E.D. 1991. Proc. Soc. Exp. Biol. Med.196, 130.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feiyan Fu et al.
Toxicology, 430, 152346-152346 (2019-12-21)
V-type agents are highly toxic organophosphorus nerve agents that inhibit acetylcholinesterase in the nervous system, causing a series of poison symptoms. Trace analytical methods are essential for the specific verification of exposure to these agents, especially for human exposure. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service