Skip to Content
Merck
All Photos(1)

Documents

235429

Sigma-Aldrich

Caspase Inhibitor Set I

The Caspase Inhibitor Set I controls the biological activity of Caspase-1. This collection of small molecule/inhibitor is primarily used for Cancer applications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

form

solid

potency

200 pM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

shipped in

ambient

storage temp.

−20°C

General description

Contains 1 mg of Caspase-3 Inhibitor II, Z-DEVD-FMK (Cat. No. 264155); 1 mg of Caspase-1 Inhibitor I, Ac-YVAD-CHO (Cat. No. 400010); and 1 mg of Caspase Inhibitor I, Z-VAD-FMK (Cat. No. 627610). Supplied with a data sheet.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
caspase-1
Reversible: no
Secondary Target
Caspase-3, Caspase-4, Caspase-5, Caspase-6, Caspase-7, Caspase-8, Caspase-10

Warning

Toxicity: Standard Handling (A)

Other Notes

Masuda, Y., et al. 1997. Biochem. Biophys. Res. Com.234, 641.
Slee, E.A., et al. 1996. Biochem. J.315, 21.
Zhu, H., et al. 1995. FEBS Lett.374, 303.
Walker, N.P.C., et al. 1994. Cell 78, 343.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service