Skip to Content
Merck
All Photos(1)

Key Documents

234374

Sigma-Aldrich

Granulocyte Macrophage Colony-Stimulating Factor, Mouse, Recombinant, E. coli

Granulocyte Macrophage Colony-Stimulating Factor, Mouse, Recombinant, E. coli, is a pleiotropic cytokine that can stimulate the proliferation, maturation, and function of hematopoietic cells.

Synonym(s):

rmGM-CSF

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$1,090.00

$1,090.00


Estimated to ship on29 May 2025



Select a Size

Change View
10 μG
$1,090.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.42

$1,090.00


Estimated to ship on29 May 2025


recombinant

expressed in E. coli

Quality Level

description

Merck USA index - 14, 4538

Assay

>97% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1.0 EU/μg Endotoxin (EU/μg GM-CSF)

shipped in

ambient

storage temp.

−70°C

General description

Recombinant, mouse granulocyte macrophage colony-stimulating factor expressed in E. coli. Pleiotropic cytokine that stimulates the proliferation, maturation, and function of hematopoietic cells. Reported to improve the response rate to antibiotic therapy in cancer subjects. Activates JAK2 in a variety of cells. Target cells include macrophages, granulocytes, and eosinophils. GM-CSF is produced by many cell types, including endothelial cells, fibroblasts, and T-lymphocytes. It can be extracted from most tissues in concentrations exceeding that of serum levels. While many cells produce GM-CSF constitutively, levels of this lymphokine increase rapidly in response to endotoxin, phorbol esters, and certain antigens.

Biochem/physiol Actions

ED₅₀ = 100-300 pg/ml as measured in a cell proliferation assay using a factor-dependent cell line, DA-3

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from sterile-filtered PBS containing 50 µg BSA/µg GM-CSF.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C) for long term storage or refrigerate (4°C) for short term storage. Avoid freeze/thaw cycles of solutions. Stock solutions are stable for 3 months at 4°C.
Reconstitute to a concentration of ≥10 µg/ml with sterile PBS containing ≥0.1% HSA or BSA.

Other Notes

Anaissie, E.J., et al. 1996. Am. J. Med. 100, 15.
Watanabe, S., et al. 1996. J. Biol. Chem.271, 12681.
Moore, M.A. 1991. Annu. Rev. Immunol. 9, 159.
Nicola, N.A. 1989. Annu. Rev. Biochem. 58, 45.
Gough, N.M., et al. 1985. EMBO J. 4, 645.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service