Skip to Content
Merck
All Photos(1)

Key Documents

233526

Sigma-Aldrich

Coagulation Factor Xa, Human Plasma

Coagulation Factor Xa, Human Plasma, CAS 9002-05-5, is a native coagulation factor Xa prepared from Factor X by activation with Russel′s viper venom. The venom is removed after activation.

Synonym(s):

Coagulation Factor Xa, Human Plasma

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

form

lyophilized

specific activity

≥170 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Native coagulation factor Xa from human plasma prepared from homogeneous Factor X by activation with Russel’s viper venom. Hydrolyzes peptide and ester bonds at the carboxyl group of arginine. Specifically catalyzes the activation of prothrombin to thrombin. Prothrombin activation is accelerated by Factor Va, phospholipids, and Ca2+. Factor Xa was prepared from homogeneous Factor X, using Russels′ viper venom (RVV), which was removed after activation. Inhibited by AEBSF (Cat. No. 101500), DFP (Cat. No. 30967), PMSF (Cat. No. 52332), and soybean trypsin inhibitor (Cat. No. 650357). Sold on the basis of plasma equivalent units of Factor X. 100 plasma equivalent units are equal to 800 µg of Factor X.
Native coagulation factor Xa from human plasma prepared from homogeneous Factor X by activation with Russel’s viper venom. The venom is removed after activation. During coagulation, in the presence of Ca2+ and membrane phospholipids, a complex of Factor Xa and Factor Va (the prothrombinase complex) activates prothrombin to thrombin. Inhibitors include AEBSF (Cat. No. 101500), DFP (Cat. No. 30967), PMSF (Cat. No. 52332), and Soybean Trypsin Inhibitor (Cat. No. 650357). Sold on the basis of plasma equivalent units of Factor X. 100 plasma equivalent units are equal to 800 µg of Factor X.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol CHROMOZYM® X substrate per min at 25°C.

Physical form

Lyophilized from 700 mM NaCl, 20 mM Tris-HCl, pH 7.4.

Analysis Note

homogeneous by SDS-PAGE

Other Notes

Heeb, M.J., et al. 1994. Proc. Natl. Acad. Sci. USA91, 2728.
Lindhout, T., et al. 1994. Biochem. J.297, 131.
Morishita, H., et al. 1994. Thrombosis Res.73, 193.
Lupu, C., et al. 1993. Thrombosis Haemostasis70, 579.
Jordan, S.P., et al. 1992. Biochemistry31, 5374.
Schoen, P., et al. 1992. Br. J. Haemotol.81, 255.
Tracy, P.B., et al. 1992. Methods Enzymol.215, 329.
Lawson, J.H., and Mann, K.G. 1991. J. Biol. Chem.226, 11317.
Tans, G., et al. 1991. J. Biol. Chem.266, 21864.
Owen, W.G., et al. 1974. J. Biol. Chem.249, 594.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Chromozym is a registered trademark of Pentapharm AG, Basel

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service