Skip to Content
Merck
All Photos(1)

Key Documents

233480

Sigma-Aldrich

Coagulation Factor XI, Human Plasma

Coagulation Factor XI, Human Plasma, CAS 9013-55-2, is a native coagulation factor XI with two identical polypeptide chains linked by disulfide bond. It is activated to Factor XIa by Factor XIIa.

Synonym(s):

Coagulation Factor XI, Human Plasma

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$903.00

$903.00


Estimated to ship on28 May 2025



Select a Size

Change View
100 μG
$903.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

$903.00


Estimated to ship on28 May 2025


Quality Level

description

Merck USA index - 14, 3924

form

liquid

specific activity

≥170 units/mg protein (clotting assay)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Coagulation Factor XI, Human Plasma, CAS 9013-55-2, is a native coagulation factor XI with two identical polypeptide chains linked by disulfide bond. It is activated to Factor XIa by Factor XIIa.
Native coagulation factor XI from human plasma. Consists of two identical polypeptide chains linked by disulfide bonds, that yield a single band at 80,000 on reduction with β-mercaptoethanol. In vivo, Factor XI is activated to Factor XIa by Factor XIIa.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 4 mM sodium acetate buffer, pH 5.3.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Analysis Note

Clotting activity is determined by comparison to a standard curve produced using normal pooled human plasma
single band by SDS-PAGE

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service