Skip to Content
Merck
All Photos(1)

Key Documents

231085

Sigma-Aldrich

Cilostamide

A cell-permeable selective inhibitor of cGMP-inhibited phosphodiesterase (PDE III; IC₅₀ = 70 nM).

Synonym(s):

Cilostamide, N-Cyclohexyl-N-methyl-4-(1,2-dihydro-2-oxo-6-quinolyloxy)butyramide, OPC 3689

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$409.00

$409.00


Estimated to ship on21 May 2025



Select a Size

Change View
10 MG
$409.00

About This Item

Empirical Formula (Hill Notation):
C20H26N2O3
CAS Number:
Molecular Weight:
342.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$409.00


Estimated to ship on21 May 2025


Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[nH]1c2c(cc[c]1=O)cc(cc2)OCCCC(=O)N(C3CCCCC3)C

InChI

1S/C20H26N2O3/c1-22(16-6-3-2-4-7-16)20(24)8-5-13-25-17-10-11-18-15(14-17)9-12-19(23)21-18/h9-12,14,16H,2-8,13H2,1H3,(H,21,23)

InChI key

UIAYVIIHMORPSJ-UHFFFAOYSA-N

General description

A cell-permeable selective inhibitor of cGMP-inhibited phosphodiesterase (PDE III; IC50 = 70 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PDE 3
Product does not compete with ATP.
Reversible: no
Target IC50: 70 nM against PDE III

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 6 months at 4°C.

Other Notes

Verghese, M.W., et al. 1995. J. Pharmacol. Exp. Ther. 272, 1313.
Christensen, S.B., and Trophy, T.J. 1994. Ann. Rep. Med. Chem. 29, 185.
Tang, K.M., et al. 1994. Eur. J. Pharmacol. 268, 105.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natasja G J Costermans et al.
Journal of visualized experiments : JoVE, (172) (2021-07-06)
Events associated with oocyte nuclear maturation have been well described. However, much less is known about the molecular pathways and processes that take place in the cytoplasm in preparation for fertilization and acquisition of totipotency. During oocyte maturation, changes in
Chisato Kunitomi et al.
Development (Cambridge, England), 151(11) (2024-05-24)
The RNA-binding protein cytoplasmic polyadenylation element binding 1 (CPEB1) plays a fundamental role in regulating mRNA translation in oocytes. However, the specifics of how and which protein kinase cascades modulate CPEB1 activity are still controversial. Using genetic and pharmacological tools
Leen Vanhoutte et al.
Human reproduction (Oxford, England), 22(5), 1239-1246 (2007-02-17)
The use of hormones for controlled ovarian stimulation results in follicular heterogeneity, with oocytes at diverse stages of nuclear and cytoplasmic development. This study evaluated the impact of temporary nuclear arrest by a specific phosphodiesterase 3-inhibitor (PDE3-I), cilostamide, on nuclear
Line M Grønning et al.
FEBS letters, 580(17), 4126-4130 (2006-07-11)
Overexpression of forkhead transcription factor FOXC2 in white adipose tissue (WAT) leads to a lean phenotype resistant to diet-induced obesity. This is due, in part, to enhanced catecholamine-induced cAMP-PKA signaling in FOXC2 transgenic mice. Here we show that rolipram treatment
Xuan G Luong et al.
Nucleic acids research, 48(6), 3257-3276 (2020-01-24)
During oocyte maturation, changes in gene expression depend exclusively on translation and degradation of maternal mRNAs rather than transcription. Execution of this translation program is essential for assembling the molecular machinery required for meiotic progression, fertilization, and embryo development. With

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service