Skip to Content
Merck
All Photos(1)

Key Documents

230914

Sigma-Aldrich

Chymotrypsin Substrate II, Fluorogenic

Sensitive fluorogenic substrate for the quantitative determination of chymotrypsin activity.

Synonym(s):

Chymotrypsin Substrate II, Fluorogenic, Suc-AAPF-AMC

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$174.00

$174.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$174.00

About This Item

Empirical Formula (Hill Notation):
C34H39N5O9
CAS Number:
Molecular Weight:
661.70
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$174.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

DMSO: soluble
acetonitrile: water (1:1): soluble

fluorescence

λex ~380 nm
λem ~460 nm

shipped in

ambient

storage temp.

−20°C

SMILES string

N1([C@@H](CCC1)C(=O)NC(=O)[C@@H](Nc3cc4c(cc3)C(=CC(=O)O4)C)Cc2ccccc2)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)CCC(=O)O)C)C

InChI

1S/C34H39N5O9/c1-19-16-30(43)48-27-18-23(11-12-24(19)27)37-25(17-22-8-5-4-6-9-22)32(45)38-33(46)26-10-7-15-39(26)34(47)21(3)36-31(44)20(2)35-28(40)13-14-29(41)42/h4-6,8-9,11-12,16,18,20-21,25-26,37H,7,10,13-15,17H2,1-3H3,(H,35,40)(H,36,44)(H,41,42)(H,38,45,46)/t20-,21-,25-,26-/m0/s1

InChI key

IGKPQFYMWKQDQS-KCXKOMAXSA-N

General description

Sensitive fluorogenic substrate for the quantitative determination of chymotrypsin activity (excitation maximum: ~380 nm; emission maximum: ~460 nm).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Chymotrypsin
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Suc-Ala-Ala-Pro-Phe-AMC

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Irvine, G.B., et al. 1990. Anal. Biochem.185, 304.
Graf, L., et al. 1987. Biochemistry26, 2616.
Oshima, G. 1984. J. Biochem.95, 1131.
Oshima, G. 1983. J. Biochem.94, 1615.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service