Skip to Content
Merck
All Photos(1)

Key Documents

219465

Sigma-Aldrich

Celastrol, Celastrus scandens

A cell-permeable dienone-phenolic triterpene compound that exhibits antioxidant and anti-inflammatory properties.

Synonym(s):

Celastrol, Celastrus scandens , Tripterin, 3-Hydroxy-24-nor-2-oxo-1(10),3,5,7-friedelatetraen-29-oic Acid, Proteasome Inhibitor XIX

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$338.00

$338.00


Estimated to ship on25 May 2025



Select a Size

Change View
10 MG
$338.00

About This Item

Empirical Formula (Hill Notation):
C29H38O4
CAS Number:
Molecular Weight:
450.61
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$338.00


Estimated to ship on25 May 2025


Quality Level

Assay

≥95% (TLC)

form

crystals

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red

solubility

DMSO: 5 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C29H38O4/c1-17-18-7-8-21-27(4,19(18)15-20(30)23(17)31)12-14-29(6)22-16-26(3,24(32)33)10-9-25(22,2)11-13-28(21,29)5/h7-8,15,22,31H,9-14,16H2,1-6H3,(H,32,33)/t22-,25-,26-,27+,28-,29+/m1/s1

InChI key

KQJSQWZMSAGSHN-JJWQIEBTSA-N

General description

A cell-permeable dienone-phenolic triterpene compound that exhibits antioxidant and anti-inflammatory properties. It suppresses LPS-induced pro-inflammatory cytokines release in macrophages and monocytes (IC50 = 40 nM for IL-1β and IL-1α, 80 nM for IL-6, 110 nM for PGE2, and 210 nM for IL-8 and TNF-α), as well as LPS-induced NF-κB activation (IC50 = 270 nM) and NO production (IC50 = 230 nM) in RAW264.7 cells. Preferentially inhibits chymotrypsin-like activity of 20S proteasome with an IC50 of 2.5 µM. Has also been shown to inhibit lipid peroxidation induced by ADP and Fe2+ in rat liver mitochondria (IC50 = 7 µM). Significantly improves the performance in memory, learning, and psychomotor activity in rats. May be useful to minimize inflammation in Alzheimer′s disease (AD).

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no
Target IC50: 2.5 µM inhibiting chymotrypsin-like activity of 20S proteasome

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Yang, H., et al. 2006. Cancer Res.66, 4758.
Westerheide, S.D., et al. 2004. J. Biol. Chem.279, 56053.
Jin, H.Z., et al. 2002. J. Nat. Prod.65, 89.
Allison, A.C., et al. 2001. Prog. Neuropsychopharmacol. Biol. Psychiatry25, 1341.
He, W., et al. 1998. Bioorg. Med. Chem. Lett.8, 3659.
Sassa, H., et al. 1990. Biochem. Biophys. Res. Commun.172, 890.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Akram Hossain et al.
Frontiers in cell and developmental biology, 9, 738502-738502 (2021-11-20)
The maintenance of genome integrity and fidelity is vital for the proper function and survival of all organisms. Recent studies have revealed that APE2 is required to activate an ATR-Chk1 DNA damage response (DDR) pathway in response to oxidative stress
Aoi Nakazono et al.
The Journal of biological chemistry, 293(38), 14758-14774 (2018-08-11)
Amyloid and amyloid-like protein aggregations are hallmarks of multiple, varied neurodegenerative disorders, including Alzheimer's and Parkinson's diseases. We previously reported that spinocerebellar ataxia type 14 (SCA14), a dominant-inherited neurodegenerative disease that affects cerebellar Purkinje cells, is characterized by the intracellular

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service