Skip to Content
Merck
All Photos(1)

Key Documents

218799

Sigma-Aldrich

Caspase-6, Human, Recombinant, E. coli

Caspase-6, Human, Recombinant is expressed as a single polypeptide in E. coli. Fused at the N- and C- termini to a His•Tag sequence. Undergoes spontaneous autoprocessing to yield the active enzyme.

Synonym(s):

Mch2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Product 218799 is not currently sold in your country. Contact Technical Service

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized

specific activity

≥6000 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

M.W. 11,000 and 18,000 (heterodimer).
Recombinant, human caspase-6 fused at the N- and C- termini to a His•Tag sequence and expressed in E. coli as a single polypeptide. Subsequently undergoes spontaneous autoprocessing to yield the active enzyme. Useful for the study of enzyme regulation and kinetics, cleavage of target substrates, and inhibitor screening. MW: 11000 and 18000.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will cleave 1 nmol Ac-VEID-pNA (Cat. No. 218762) per h at 37°C, pH 7.2.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 3 months at -70°C.
Reconstitute to 1 unit/µl in PBS, 15% glycerol.

Other Notes

LeBlanc, A., et al. 1999. J. Biol. Chem.274, 23426.
Stennicke, H.R., and Salvesen, G.S. 1997. J. Biol. Chem.272, 25719.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunyan Wang et al.
Neuroscience letters, 721, 134802-134802 (2020-02-06)
Neuroinflammation in the spinal cord is a pathological event in remifentanil-induced hyperalgesia (RIH), but its underlying molecular mechanisms remain unclear. Recent studies recapitulate the significance of the intracellular protease caspase-6 in the release of inflammatory mediators and synaptic plasticity in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service