Skip to Content
Merck
All Photos(1)

Key Documents

211275

Sigma-Aldrich

Capsaicin

An active constituent of cayenne pepper with excitatory and desensitizing effects on a subset of primary afferent sensory neurons.

Synonym(s):

Capsaicin, [(E)-N-(4-Hydroxy-3-methoxyphenyl)methyl]-8-methyl-6-nonenamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$188.00

$188.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$188.00

About This Item

Empirical Formula (Hill Notation):
C18H27NO3
CAS Number:
Molecular Weight:
305.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$188.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 1768

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

ethanol: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

N(Cc1cc(c(cc1)O)OC)C(=O)CCCC\C=C\C(C)C

InChI

1S/C18H27NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h6,8,10-12,14,20H,4-5,7,9,13H2,1-3H3,(H,19,21)/b8-6+

InChI key

YKPUWZUDDOIDPM-SOFGYWHQSA-N

General description

An active constituent of cayenne pepper with excitatory and desensitizing effects on a subset of primary afferent sensory neurons. An antagonist of acetylcholine-induced responses in rat PC12 cells. Exhibits anti-nociceptive and anti-inflammatory effects. Inhibits NF-κB activation by TNF. Reversibly inhibits platelet aggregation.
An active constituent of cayenne pepper. Has excitatory and desensitizing effects on a subset of primary afferent sensory neurons. An antagonist of acetylcholine-induced respones in rat pheochromocytoma cells. Exhibits antinociceptive and anti-inflammatory effects. Inhibits NF-κB activation by TNF. Reversibly inhibits platelet aggregation.

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Singh, S., et al. 1996. J. Immunol.157, 4412.
Nakazawa, K., et al. 1994. Br. J. Pharmacol.113, 296.
Simmons, D.D., et al. 1994. Int. J. Dev. Neurosci.12, 517.
Takaki, M., et al. 1994. J. Mol. Cell. Cardiol.26, 1227.
Bevan, S., and Szolcsanyi, J. 1990. Trends Pharmacol. Sci.11, 330.
Pernow, B., et al. 1985. J. Immunol.135, 8125.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Gao et al.
Diabetes, 69(2), 165-180 (2019-11-13)
The whitening and loss of brown adipose tissue (BAT) during obesity and aging promote metabolic disorders and related diseases. The imbalance of Ca2+ homeostasis accounts for the dysfunction and clearance of mitochondria during BAT whitening. Capsaicin, a dietary factor activating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service