Skip to Content
Merck
All Photos(1)

Key Documents

208771

Sigma-Aldrich

Calpain Substrate III, Fluorogenic

A fluorogenic FRET peptide with a substrate sequence that is optimized for calpain-1 and -2.

Synonym(s):

Calpain Substrate III, Fluorogenic, (DABCYL)-TPLK~SPPPSPR-(EDANS)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$441.00

$441.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$441.00

About This Item

Empirical Formula (Hill Notation):
C80H114N20O18S
Molecular Weight:
1675.95
UNSPSC Code:
12352200
NACRES:
NA.77

$441.00


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

fluorescence

λex 320 nm
λem 480 nm

shipped in

wet ice

storage temp.

−20°C

General description

A fluorogenic FRET peptide with a substrate sequence that is optimized for calpain-1 and -2 based on 106 known cleavage sites. It is cleaved by mammalian calpains with higher efficiency (kcat/Kmx10-2 (M-1s-1) = 69.8, 38.5, 20.0, 6.81, 3.58, 0.05, and <0.05 for calpain-2, trypsin, papain, calpain-B, calpain-A, chymotrypsin, and cathepsin-B, respectively). It is kinetically superior to other commonly used calpain substrates, such as LY-AMC (Cat. No. 208731) and α-spectrin cleavage site-based substrate (Cat. No. 208748). Has been used successfully in monitoring calpain activity in intact Drosophila S2 cells (cellular incorporation achieved with a lipofection reagent) and in COS-7 cell lysate.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Substrate sequence that is optimized for calpain-1 and -2
Product does not compete with ATP.
Reversible: no
kcat/Kmx10-2 (M-1s-1) = 69.8, 38.5, 20.0, 6.81, 3.58, 0.05, and <0.05 for calpain-2, trypsin, papain, calpain-B, calpain-A, chymotrypsin, and cathepsin-B, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Sequence

DABCYL-Thr-Pro-Leu-Lys~Ser-Pro-Pro-Pro-Ser-Pro-Arg-EDANS

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tompa, P., et al. 2004. J. Biol. Chem.279, 20775.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service