Skip to Content
Merck
All Photos(1)

Key Documents

208743

Sigma-Aldrich

Calpain Inhibitor XI

The Calpain Inhibitor XI, also referenced under CAS 145731-49-3, controls the biological activity of Calpain. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Calpain Inhibitor XI, Z-L-Abu-CONH(CH₂)₃-morpholine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$876.00

$876.00


Estimated to ship on29 May 2025



Select a Size

Change View
5 MG
$876.00

About This Item

Empirical Formula (Hill Notation):
C26H40N4O6
CAS Number:
Molecular Weight:
504.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$876.00


Estimated to ship on29 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N2(CCOCC2)CCCNC(=O)C(=O)C(NC(=O)C(NC(=O)OCc1ccccc1)CC(C)C)CC

InChI

1S/C26H40N4O6/c1-4-21(23(31)25(33)27-11-8-12-30-13-15-35-16-14-30)28-24(32)22(17-19(2)3)29-26(34)36-18-20-9-6-5-7-10-20/h5-7,9-10,19,21-22H,4,8,11-18H2,1-3H3,(H,27,33)(H,28,32)(H,29,34)

InChI key

TZVQRMYLYQNBOA-UHFFFAOYSA-N

General description

A cell-permeable dipeptidyl α-ketoamide that acts as a potent, highly selective, reversible, and active site inhibitor of calpain-1 and -2 (Ki = 140 nM and 41 nM, respectively). Weakly inhibits cathepsin B (Ki = 6.9 µM). Reported to have a neuroprotective role in the central nervous system following focal ischemia. Also protects against virus-induced apoptotic myocardial injury in mice.
A cell-permeable dipeptidyl a-ketoamide that acts as a potent, highly selective, reversible, active site inhibitor of calpain-1 (Ki = 140 nM) and calpain-2 (Ki = 41 nM). Weakly inhibits cathepsin B (Ki = 6.9 µM). Shown to inhibit calpain-m-mediated degradation of neurofilament protein (NFP) (IC50 = 600 nM). Also shown to exhibit neuroprotective effects in the central nervous system following focal ischemia. Reported to protect against reovirus-induced myocarditis in mice.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
calpain 1, calpain 2
Product does not compete with ATP.
Reversible: yes
Target Ki: 140 nM and 41 nM, against calpain-1 and -2, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Leu-Abu-CONH(CH₂)₃-morpholine (Abu = α-aminobutyric acid)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Blomgren, K., et al. 2001. J. Biol. Chem.276, 10191.
DeBiasi, R.L., et al. 2001. J. Virol.75, 351.
Saatman, K.E., et al. 2000. J. Cereb. Blood Flow Metab.20, 66.
Stelmasiak, Z., et al. 2000. Med. Sci. Monit.6, 426.
Blomgren, K., et al. 1999. J. Biol. Chem.274, 14046.
James, T., et al. 1998. J. Neurosci. Res.51, 218.
Li, Z., et al. 1996. J. Med. Chem.39, 4089.
Saatman, K.E, et al. 1996. Proc. Natl. Acad. Sci. USA93, 3428.
Bartus, R.T., et al. 1995. Neurol. Res.17, 249.
Bartus, R.T., et al. 1994. Stroke25, 2265.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service