Skip to Content
Merck
All Photos(1)

Key Documents

204902

Sigma-Aldrich

Complement C5a des-Arg, Human

Complement C5a des-Arg, Human, is a native, single chain glycopeptide (73 aa). Once produced, C5a is converted to C5a des-Arg upon removal of the C-terminal arginine by serum carboxypeptidase N.

Synonym(s):

C5a Complement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Native, human des-Arg C5a complement component. Single chain glycopeptide composed of 73 amino acids. Once the C5a glycopeptide has been produced in human plasma or serum, it is rapidly converted to C5a des-Arg upon removal of the C-terminal arginine by the endogenous serum carboxypeptidase N. Unlike the C3a des-Arg peptide, C5a des-Arg retains significant levels of its biological activities. Thus, C5a des-Arg has been shown to exhibit inflammatory cell chemotaxis, smooth muscle contraction, and leukotriene release from guinea pig lung.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 120 mM NaCl, 10 mM HEPES, pH 7.2.

Preparation Note

Prepared from plasma of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Webster, R.O., et al. 1980. Immunopharmacology 1, 201.
McCarthy, K. and Henson, P.M. 1979. J. Immunol. 123, 2511.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service