Skip to Content
Merck
All Photos(1)

Key Documents

203895

Sigma-Aldrich

BTS

A potent inhibitor of Ca2+-stimulated myosin S1 ATPase (IC₅₀ = ~ 5 µM) and reversibly blocks the gliding motility.

Synonym(s):

BTS, N-Benzyl- p-toluenesulphonamide, N-Benzyl-p-toluenesulphonamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$247.00

$247.00


Estimated to ship on16 April 2025



Select a Size

Change View
5 MG
$247.00

About This Item

Empirical Formula (Hill Notation):
C14H15NO2S
CAS Number:
Molecular Weight:
261.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$247.00


Estimated to ship on16 April 2025


Quality Level

description

RTECS - XT5490000

Assay

≥95% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NCc2ccccc2)c1ccc(cc1)C

InChI

1S/C14H15NO2S/c1-12-7-9-14(10-8-12)18(16,17)15-11-13-5-3-2-4-6-13/h2-10,15H,11H2,1H3

InChI key

WTHKAJZQYNKTCJ-UHFFFAOYSA-N

General description

A potent inhibitor of Ca2+-stimulated myosin S1 ATPase (IC50 = ~ 5 µM) and reversibly blocks the gliding motility. It also weakens myosin’s interaction with F-actin. Much less effective in suppressing contraction in rat myocardial or rabbit slow twitch muscle and has no effect on platelet myosin II.
An aryl sulphonamide that acts as a potent inhibitor of Ca2+-stimulated myosin S1 actin-stimulated ATPase activity (IC50 = ~5 µM). Also blocks actin-stimulated ATPase activity with similar potency (IC50 = 5 µM). Reversibly blocks gliding motility of skeletal muscle myosin (IC50<2 µM). Less effective in suppressing contraction in rat myocardial and rabbit slow twitch muscle compared to skeletal muscle. Does not inhibit platelet myosin II.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Ca2+-stimulated myosin S1 ATPase
Product does not compete with ATP.
Reversible: yes
Target IC50: ~5 µM against Ca2+-stimulated myosin S1 ATPase

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Cheung, A., et al. 2002. Nat. Cell Biol.4, 83.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service