Skip to Content
Merck
All Photos(1)

Key Documents

20-309

Sigma-Aldrich

0.1M MnCl2

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
$386.00

$386.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
500 μL
$386.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41

$386.00


Estimated to ship on21 May 2025


Request a Bulk Order

form

liquid

Quality Level

manufacturer/tradename

Upstate®

shipped in

wet ice

Application

Research Category
All

Storage and Stability

stable 6 months at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aminopeptidase N from Bombyx mori as a candidate for the receptor of Bacillus thuringiensis Cry1Aa toxin.
K Yaoi,T Kadotani,H Kuwana,A Shinkawa,T Takahashi,H Iwahana,R Sato
European Journal of Biochemistry null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service