Skip to Content
Merck
All Photos(1)

Key Documents

1.99041

Supelco

Buffer solutions

(boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 11.00 (25°C) Certipur®

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$38.80
4 L
$270.00

$38.80


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$38.80
4 L
$270.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.25

$38.80


Estimated to ship on31 May 2025


Request a Bulk Order

grade

certified reference material

Quality Level

product line

Certipur®

pH

11.0 (25 °C in H2O)

density

1.01 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The boric acid/ potassium chloride/sodium hydroxide buffer solution is gravimetrically prepared at a registered calibration laboratory with high-quality reagents and water. Its pH value of 11.00 at 25°C is measured using a combined glass electrode following a 5-point calibration method. The measurement is made following DIN 19268 using reference buffer solutions compliant to DIN 19266, NIST, IUPAC, Ph. Eur, and USP. The buffer pH is temperature dependent and therefore a constant temperature must be maintained during its use.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

The buffer is used as a standard to calibrate pH measuring instruments or pH electrodes and as a control to measure the pH of other solutions.

Features and Benefits

  • Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to secondary reference standard from NIST and primary certified reference standard from PTB
  • Available in a secure 500 mL PE bottle and 4 L innovative and sustainable Titripac® PE/Aluminum packaging for a reduced waste generation
  • Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
  • Product label follows the ISO Guide 31 and contains information about ordering number, article description, lot number, safety data, and expiry date
  • Packaging material subjected to comprehensive stability studies as per ISO Guide 35

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripac is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service