1.93678
Mercury(II) iodide red
EMPLURA®
Synonym(s):
Mercury(II) iodide red,
About This Item
Recommended Products
product line
EMPLURA®
storage temp.
room temp
SMILES string
[Hg+2].[I-].[I-]
InChI
1S/Hg.2HI/h;2*1H/q+2;;/p-2
InChI key
YFDLHELOZYVNJE-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Legal Information
Disclaimer
![](/deepweb/assets/sigmaaldrich/product/images/242/317/89e134be-e488-4e6d-9500-3baa2929e5c3/300/89e134be-e488-4e6d-9500-3baa2929e5c3.jpg)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service