Skip to Content
Merck
All Photos(2)

Key Documents

191500

Millipore

1-Azakenpaullone

A Kenpaullone analog that acts as a potent and ATP-competitive inhibitor of GSK-3β.

Synonym(s):

1-Azakenpaullone, 1-AKP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10BrN3O
CAS Number:
Molecular Weight:
328.16
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥94% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C15H10BrN3O/c16-8-3-4-11-9(6-8)10-7-13(20)18-12-2-1-5-17-15(12)14(10)19-11/h1-6,19H,7H2,(H,18,20)

InChI key

NTSBZVCEIVPKBJ-UHFFFAOYSA-N

General description

A Kenpaullone (Cat. No. 422000) analog that acts as a potent and ATP-competitive inhibitor of GSK-3β (IC50 = 18 nM), and displays ~100-200 fold greater selectivity over Cdk1/B and Cdk5/p25 (IC50 = 2 µM and 4.2 µM, respectively). Also available as a 10 mM solution in DMSO (Cat. No. 191501).
A Kenpaullone (Cat. No. 422000) analog that acts as a potent and ATP-competitive inhibitor of GSK-3β (IC50 = 18 nM). Displays over 100-fold greater selectivity over Cdk1/cyclin B and Cdk5/p25 (IC50 = 2.0 µM and 4.2 µM, respectively).

Biochem/physiol Actions

Cell permeable: no
Primary Target
GSK-3β
Product competes with ATP.
Reversible: no
Target IC50: 18 nM against GSK-3β

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kunick, C., et al. 2004. Bioorg. Med. Chem. Lett.14, 413.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service