Skip to Content
Merck
All Photos(1)

Key Documents

19-143

Sigma-Aldrich

PD 98059

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$307.00

$307.00


Estimated to ship on27 May 2025



Select a Size

Change View
5 MG
$307.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

$307.00


Estimated to ship on27 May 2025


form

solid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

shipped in

wet ice

Application

Potent, selective and cell-permeable inhibitor of MAPK kinase, MAPK/ERK, or MEKK.

Biochem/physiol Actions

Inhibitor Type: Kinase

Physical form

C16H13NO3

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Liu et al.
Non-coding RNA research, 9(3), 901-912 (2024-04-15)
DNA methylation is a crucial epigenetic alteration involved in diverse biological processes and diseases. Nevertheless, the precise role of DNA methylation in chemotherapeutic drug-induced alopecia remains unclear. This study examined the role and novel processes of DNA methylation in regulating
Shang Li et al.
NPJ biofilms and microbiomes, 9(1), 47-47 (2023-07-09)
Knowledge gaps that limit the development of therapies for polycystic ovary syndrome (PCOS) concern various environmental factors that impact clinical characteristics. Circadian dysrhythmia contributes to glycometabolic and reproductive hallmarks of PCOS. Here, we illustrated the amelioration of Limosilactobacillus reuteri (L.
D R Alessi et al.
The Journal of biological chemistry, 270(46), 27489-27494 (1995-11-17)
PD 098059 has been shown previously to inhibit the dephosphorylated form of mitogen-activated protein kinase kinase-1 (MAPKK1) and a mutant MAPKK1(S217E,S221E), which has low levels of constitutive activity (Dudley, D. T., Pang, L., Decker, S. J., Bridges, A. J., and
Qize Wei et al.
Molecular biology of the cell, 13(2), 683-697 (2002-02-21)
We ectopically expressed the transcription factor Pitx2a, one of the Pitx2 isoforms, in HeLa cells by using a tetracycline-inducible expression system and examined whether Pitx2a was capable of modulating Rho GTPase signaling and altering the cell's cytoskeleton. Ectopic expression of
L Pang et al.
The Journal of biological chemistry, 270(23), 13585-13588 (1995-06-09)
The mitogen-activated protein kinase (MAP kinase) pathway is thought to play an important role in the actions of neurotrophins. A small molecule inhibitor of the upstream kinase activator of MAP kinase, MAP kinase kinase (MEK) was examined for its effect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service