Skip to Content
Merck
All Photos(1)

Key Documents

1831

Millipore

L-Ascorbic Acid, Free Acid

Protects cells against damaging effects of radiation and oxygen radicals. Increases rate of mineralization in osteoblasts.

Synonym(s):

L-Ascorbic Acid, Free Acid, Vitamin C

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$346.00

$346.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
1 KG
$346.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.21

$346.00


In StockDetails


Request a Bulk Order

grade

for cell biology

description

RTECS - CI7650000
Merck USA index - 14, 830

Assay

≥99% (by assay)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

cell culture | mammalian: suitable

color

white to light yellow

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-8,10-11H,1H2

InChI key

TYQCGQRIZGCHNB-UHFFFAOYSA-N

General description

Protects cells against damaging effects of radiation and oxygen radicals. Increases rate of mineralization in osteoblasts. A cofactor involved in hydroxylation of proline and lysine. Reduces Na+ - Ca2+ exchange in cultured astrocytes. Modulates cyclic nucleotide levels in B and T cells. Inhibits apoptosis in cultured rat ovarian follicles.
Protects cells against the damaging effects of radiation and oxygen radicals. Increases the rate of mineralization in osteoblasts. A cofactor involved in hydroxylation of proline and lysine. Reduces Na+-Ca2+ exchange in cultured astrocytes. Modulates cyclic nucleotide levels in B and T cells. Inhibits apoptosis in cultured rat ovarian follicles.

Application


  • The comparison of contribution of GO and rGO produced by green synthesis to the properties of CMC-based wound dressing material.: This study evaluates the enhancement of wound dressing materials through the incorporation of graphene oxide (GO) and reduced graphene oxide (rGO) synthesized via green methods, emphasizing the role of L-Ascorbic Acid, Free Acid in stabilizing these nanomaterials. This research demonstrates the compound′s critical role in biomedical applications, particularly in enhancing the biocompatibility and mechanical properties of wound dressings (Karaca ÖG et al., 2024).

  • Depletion of Free Chlorine and Generation of Trichloromethane in the Presence of pH Control Agents in Chlorinated Water at pH 6.5.: Explores the environmental and health impacts of chlorination by-products where L-Ascorbic Acid, Free Acid is used to mitigate harmful reactions. This research is crucial for improving water treatment processes, ensuring safety and sustainability (Fan X et al., 2024).

  • Nutritional Quality of Conventional, Organic, and Hydroponic Tomatoes Commercialized in Quito, Ecuador.: This study assesses the nutritional content of tomatoes grown under different agricultural practices, with L-Ascorbic Acid, Free Acid evaluated for its impact on enhancing nutrient profiles. The findings contribute to the ongoing debate on the nutritional benefits of various farming methodologies (Vélez-Terreros PY et al., 2024).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Material readily oxidizes in aqueous solution; reconstitute just prior to use.

Other Notes

Takuma, K., et al. 1995. J. Neurochem. 64, 1536.
Tilly, J.L., and Tilly, K.I. 1995. Endocrinology136, 242.
Evans, P.J., et al. 1994. Biochem. Pharmacol. 48, 2173.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H H Lee et al.
Journal of food science and technology, 52(12), 7806-7816 (2015-11-26)
Whole grains consumption promotes health benefits, but demonstrates controversial impacts from phytic acid in meeting requirements of good health. Therefore, this study was aimed to determine the nutrient bioaccessibility and antioxidant properties of rice cultivars named "Adan" or "Bario" and
Yun Ha Hur et al.
Developmental cell, 56(3), 277-291 (2020-12-16)
It is critical that epiblast cells within blastocyst-stage embryos receive the necessary regulatory cues to remain pluripotent until the appropriate time when they are stimulated to undergo differentiation, ultimately to give rise to an entire organism. Here, we show that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service