Skip to Content
Merck
All Photos(1)

Key Documents

182518

Sigma-Aldrich

Arp2/3 Complex Inhibitor II, Inactive Control, CK-312

The Arp2/3 Complex Inhibitor II, Inactive Control, CK-312 controls the biological activity of Arp2/3. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Arp2/3 Complex Inhibitor II, Inactive Control, CK-312, 2-(3-Bromophenyl)-3-(4-ethoxyphenyl)-1,3-thiazolidin-4-one, racemic, CK-0993312

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$246.00

$246.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$246.00

About This Item

Empirical Formula (Hill Notation):
C17H16BrNO2S
Molecular Weight:
378.28
UNSPSC Code:
12352200
NACRES:
NA.28

$246.00


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable thiazolidinone compound that exhibits no Arp2/3 inhibitory activity and serves as an inactive control for CK-869 (Cat. No. 182516) in both cell-based and cell-free experiments.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Nolen, B.J., et al. 2009. Nature460, 1031.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service