Skip to Content
Merck
All Photos(1)

Key Documents

182015

Sigma-Aldrich

Cyclo(Arg-Gly-Asp-D-Phe-Val)

Potent inhibitor of cell adhesion.

Synonym(s):

Cyclo(Arg-Gly-Asp-D-Phe-Val), RGDFV Peptide, Cyclic

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$378.00

$378.00


Estimated to ship on01 June 2025



Select a Size

Change View
1 MG
$378.00

About This Item

Empirical Formula (Hill Notation):
C26H38N8O7
CAS Number:
Molecular Weight:
574.63
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

$378.00


Estimated to ship on01 June 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

1 N acetic acid: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Potent inhibitor of cell adhesion. Inhibits tumor cell adhesion to laminin and vitronectin substrates. In vivo investigations in rats showed it to be useful in the amelioration of ischemic acute renal failure in rats. Also inhibits tubular obstruction by preventing cell-cell adhesion. Inhibits the redistribution of c-Src into focal adhesions, thereby leading to impaired MAP Kinase activation.

Biochem/physiol Actions

Cell permeable: no
Primary Target
inhibits tumor cell adhesion to laminin and vitronectin substrates
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Other Notes

Shono, T., et al. 2001. Exp. Cell Res.268, 169.
Matsuno, H., et al. 1994. Circulation 90, 2203.
Noiri, E., et al. 1994. Kidney Int. 46, 1050.
Gurrath, M., et al. 1992. Eur. J. Biochem. 210, 911.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service