Skip to Content
Merck
All Photos(1)

Key Documents

1.80657

Ethanol (denatured)

85.5 % (v/v),83.5-87.5 % (v/v), for disinfection

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
bp:
78 °C (lit.)
vapor pressure:
44.6 mmHg ( 20 °C)

Product Name

Ethanol, denatured, 85.5 % (v/v), for disinfection

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

autoignition temp.

683 °F

quality

denatured

expl. lim.

19 %, 60 °F

concentration

83.5-87.5 % (v/v)
85.5 % (v/v)

impurities

12.5-16.5 % (v/v) water

color

, Hazen ≤10

refractive index

n20/D 1.3600 (lit.)

bp

78 °C (lit.)

mp

−114 °C (lit.)

density

0.835-0.845 g/mL at 20 °C

suitability

corresponds for identity (IR)

format

neat

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

for disinfection PT1/PT2: solution for hand and surface disinfection Wirkstoff/Active Substance: Ethanol (CAS 64-17-5), 81,6% (w/w), 85,5% (v/v)

Packaging

Package sizes:
  • 2.5 L in PE bottle
  • 5 L in PE bottle
  • 25 L in PE canister
  • 950 L in IBC PE

Analysis Note

denaturated with ethyl methyl ketone

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service