Skip to Content
Merck
All Photos(1)

Key Documents

178281

Sigma-Aldrich

p-APMSF, Hydrochloride

Alternative to DFP and PMSF.

Synonym(s):

p-APMSF, Hydrochloride, p-Amidinophenylmethylsulfonylfluoride, HCl

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$273.00

$273.00


Estimated to ship on31 May 2025



Select a Size

Change View
5 MG
$273.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$273.00


Estimated to ship on31 May 2025


Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

water: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

F[S](=O)(=O)Cc1ccc(cc1)\C(=N\[H])\N.Cl

InChI

1S/C8H9FN2O2S.ClH/c9-14(12,13)5-6-1-3-7(4-2-6)8(10)11;/h1-4H,5H2,(H3,10,11);1H

InChI key

KHLLRHIUKOJXLL-UHFFFAOYSA-N

General description

Alternative to DFP and PMSF. An irreversible inhibitor of trypsin-like serine proteases. Inhibits trypsin and thrombin in equimolar concentration. Acts on C1r, C1s, Factor Xa, and plasmin, requiring a 5- to 10-fold excess for complete irreversible inhibition. Inhibitory activity is approximately 1000-fold greater than that of PMSF.
An alternative to DFP and PMSF. Specific irreversible inhibitor of trypsin-like serine proteases. Inhibits trypsin and thrombin in equimolar concentrations. Acts on C1r, C1s, Factor Xa and plasmin, requiring a 5- to 10-fold excess for complete irreversible inhibition. Inhibitory activity is approximately 1000-fold greater than that of PMSF. Stability is pH dependent: t½ = 20 min at pH 6.0; 6 min at pH 7.0; 1 millisecond at pH 8.0.

Biochem/physiol Actions

Cell permeable: no
Primary Target
trypsin-like serine proteases
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Cole, T.C., et al. 1989. Biochim. Biophys. Acta 990, 254.
Laura, R., et al. 1980. Biochemistry19, 4859.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service