Skip to Content
Merck
All Photos(3)

Key Documents

17-685

Sigma-Aldrich

ChIPAb+ Phospho-Histone H3 (Ser10) - ChIP Validated Antibody and Primer Set

clone CMA312, from mouse, purified by using protein G

Synonym(s):

H3S10P, Histone H3 (phospho S10), H3 histone, family 3A

Sign Into View Organizational & Contract Pricing

Select a Size

25 ASSAYS
$908.00

$908.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
25 ASSAYS
$908.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.32

$908.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

clone

CMA312, monoclonal

purified by

using protein G

species reactivity

human, plant, vertebrates

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Phospho-Histone H3 (Ser10) set includes the Anti-phospho-Histone H3 (Ser10) antibody, a negative control antibody (purified Mouse IgG), and qPCR primers which amplify a 166 bp region within the promoter of the human GAPDH gene. The phospho-histone H3 (Ser10) and negative control antibodies are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of phospho-histone H3 (Ser10) associated chromatin.

Specificity

Recognizes histone H3, Mr 17 kDa, phosphorylated at Ser10.
The immunogen sequence is identical in a wide range of animal and plant species, so broad cross-reactivity is expected.

Immunogen

Epitope: a.a. 1-19
Immunogen was a synthetic peptide corresponding to amino acids 1-19 of histone H3, phosphorylated at Ser10.

Application

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from untreated or colcemid-treated HeLa cells (2 X 106 cell equivalents per IP) was subjected to chromatin immunoprecipitation using 2 μg of either a normal mouse IgG or Anti-phospho-Histone H3 (Ser10) antibody and the Magna ChIP G Kit (Cat. # 17-611). Successful immunoprecipitation of phospho-histone H3 (Ser10)-associated DNA fragments was verified by qPCR using Control Primers for untreated and treated chromatin samples (Please see figures). Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna G ChIP (Cat. # 17-409) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Western Blot Analysis:
Recombinant Histone H3 or acid extracts from colcemid-treated HeLa cells (1 μg) were resolved by electrophoresis, transferred to PVDF membrane and probed with 1 μg/mL anti-phospho Histone H3 (Ser10), clone CMA312. Proteins were visualized using goat anti-mouse secondary antibody conjugated to HRP and a chemiluminescence detection system (Please see figures).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Histones
This ChIPAb+ Phospho-Histone H3 (Ser10) -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

25 assays per set. Recommended use: ~2 μg antibody per chromatin immunoprecipitation (dependent upon biological context).

Quality

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from colcemid-treated HeLa cells (1 X 106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 2 µg of either a normal mouse IgG or Anti-phospho-Histone H3 (Ser10) antibody and the Magna ChIP® G Kit (Cat. # 17-611). Successful immuno-precipitation of phospho-histone H3 (Ser10) associated DNA fragments was verified by qPCR using Control Primers (Please see figures).
Please refer to the EZ-Magna ChIP G (Cat. # 17-409) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Target description

17 kDa

Physical form

Anti-phospho-Histone H3 (Ser10) (mouse monoclonal IgG, Clone CMA312). One vial containing 50 μg protein G-purified antibody in 50 μL PBS containing 0.05% sodium. Store at -20°C.

Normal Mouse IgG. Two vials containing 25 μg purified mouse IgG in 25 μL storage buffer containing 0.1% sodium azide. Store at -20°C.

Control Primers. One vial containing 75 μL of 5 μM of each primer specific for the promoter region of human GAPDH. Store at -20°C.
FOR: TAC TAG CGG TTT TAC GGG CG
REV: TCG AAC AGG AGG AGC AGA GAG CGA

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Includes negative control mouse IgG antibody and control primers specific for human GAPDH promoter.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iben Kümler et al.
BMC cancer, 19(1), 573-573 (2019-06-15)
Treatment options in metastatic breast cancer are limited. New therapies preferable with predictive biomarkers are needed. The aim of these trials was to investigate if gene copy number of the topoisomerase 1 gene was predictive of response to the topoisomerase
Genome-wide analysis of H4K5 acetylation associated with fear memory in mice.
Park, CS; Rehrauer, H; Mansuy, IM
BMC Genomics null
Grigory Krapivinsky et al.
Cell, 157(5), 1061-1072 (2014-05-27)
TRPM7 is a ubiquitous ion channel and kinase, a unique "chanzyme," required for proper early embryonic development. It conducts Zn(2+), Mg(2+), and Ca(2+) as well as monovalent cations and contains a functional serine/threonine kinase at its carboxyl terminus. Here, we
Min Nie et al.
Aging, 12(2), 1304-1321 (2020-01-27)
Aurora kinase B (AURKB) triggers the phosphorylation of serine 10 on histone H3 (H3S10ph), which is important for chromosome condensation and cytokinesis during mitosis in mammals. However, how exactly AURKB controls cell cycle and contributes to tumorigenesis as an oncoprotein
Four enzymes cooperate to displace histone H1 during the first minute of hormonal gene activation.
Vicent, GP; Nacht, AS; Font-Mateu, J; Castellano, G; Gaveglia, L; Ballare, C; Beato, M
Genes & Development null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service