Skip to Content
Merck
All Photos(1)

Key Documents

176695

Sigma-Aldrich

Angeli′s Salt

A nitroxyl anion (NO⁻) donor that spontaneously dissociates in a pH dependent first order process to liberate 0.54 moles of NO per mole of the parent compound.

Synonym(s):

Angeli’s Salt, Sodium α-Oxyhyponitrite, Disodium Diazen-1-ium-1,2,2-triolate, AS, Sodium Trioxodinitrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$132.00

$132.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$132.00

About This Item

Empirical Formula (Hill Notation):
N2O3 · 2Na
CAS Number:
Molecular Weight:
121.99
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$132.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥99%

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

alkaline solution: soluble (0.01 M NaOH)

shipped in

wet ice

storage temp.

−20°C

General description

A nitroxyl anion (NO) donor that spontaneously dissociates in a pH dependent first order process to liberate 0.54 moles of NO per mole of the parent compound. Half-life (t½) in 100 mM phosphate buffer, pH 7.4 is 2.3 minutes at 37°C. Decomposition is nearly instantaneous at pH 5.0.
A nitroxyl anion (NO-) donor that spontaneously, in the presence of oxygen and Cu2+ and Fe3+, dissociates in a pH-dependent, first-order process to liberate 0.54 moles of NO per mole of the parent compound. Half-life (t½) of NO release = 2.3 min. (in 100 mM phosphate buffer, pH 7.4 at 37°C). Induces relaxation of norepinephrine-constricted rabbit aorta (EC50 = 590 nM). Angeli′s Salt produces nitroxyl anion (NO-) that can be converted to NO in the presence of heme.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Nitroxyl anion (NO-) donor
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution in alkaline solution, store at -20°C. This product is stable for 6 months as supplied. Alkaline stock solutions are stable for up to 24 h at -20°C. Unstable in acidic or neutral pH buffers.

Other Notes

Miranda, K.M., et al. 2001. J. Biol. Chem.276, 1720.
Hrinczenko, B.W., et al. 2000. Br. J. Haematol.110, 412.
Nelli, S., et al. 2000. Br. J. Pharmacol.131, 356.
Ma, X.L., et al. 1999. Proc. Natl. Acad. Sci. USA96, 14617.
Keefer, L.K., et al. 1996. Methods Enzymol.268, 281.
Fukuto, J.M., et al. 1993. Biochem. Biophys. Res. Commun.196, 707.
Maragos, C.M., et al. 1991. J. Med. Chem.34, 3242.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service