Skip to Content
Merck
All Photos(3)

Key Documents

17-661

Sigma-Aldrich

ChIPAb+ SUZ12 - ChIP Validated Antibody and Primer Set

from mouse

Synonym(s):

SUZ12, polycomb protein SUZ12, suppressor of zeste 12 homolog

Sign Into View Organizational & Contract Pricing

Select a Size

25 ASSAYS
$960.00

$960.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
25 ASSAYS
$960.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.52

$960.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

monoclonal

species reactivity

vertebrates, rat, human, mouse

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
dot blot: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... SUZ12(23512)

Related Categories

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ SUZ12, set includes the polycomb protein SUZ12 antibody (also known as suppressor of zeste 12 protein homolog or chromatin precipitated E2F target 9 protein), a negative control antibody (purified mouse IgG), and qPCR primers which amplify a 138 bp region upstream of human HoxA2 gene. The SUZ12 and negative control antibodies are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of SUZ12 associated
chromatin.

Specificity

Not tested in other species.
Recognizes SUZ12, Mr 95 kDa.

Immunogen

SUZ12, purified antibody is made against human SUZ12.

Application

Chromatin Immunoprecipitation:
Representative lot data.
Sonicated chromatin prepared from Ntera2 cells (3 X 106 cell equivalents per IP) was subjected to chromatin immunoprecipitation using 1 µg of either a normal mouse IgG or Anti-SUZ12 antibody and the Magna ChIP G Kit (Cat. # 17-611).
Successful immunoprecipitation of SUZ12 associated DNA fragments was verified by qPCR using GAPDH promoter (negative) and HoxA2 (positive) Primers (Please see figures). Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna G ChIP (Cat. # 17-409) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Western Blot Analysis:
Representative lot data.
Lysates from HeLa cells were resolved by electrophoresis, transferred to PVDF and probed with anti-SUZ12 (1:1,000 dilution). Proteins were visualized using goat anti-mouse secondary antibody conjugated to HRP and chemiluminescence detection (Please see figures).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This ChIPAb+ SUZ12 -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

25 assays per set. Recommended use: ~2 μg antibody per chromatin immunoprecipitation (dependent upon biological context).

Quality

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from Ntera2 cells (3 X 106 cell equivalents per IP) were subjected to chromatin immunoprecipitation using 2 µg of either a normal mouse IgG or Anti-SUZ12 antibody and the Magna ChIP® G Kit (Cat. # 17-611).
Successful immunoprecipitation of SUZ12 associated DNA fragments was verified by qPCR using Control Primers (Please see figures).
Please refer to the EZ-Magna ChIP G (Cat. # 17-409) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Target description

~95 kDa

Physical form

Anti-SUZ12 (mouse monoclonal IgG). One vial containing 50 μg of protein A purified antibody in PBS containing 0.05% sodium azide. May contain 30% glycerol (see certificate of analysis). Store at -20°C.

Normal Mouse IgG. Two vials containing 25 μg of purified mouse IgG in 25 μL of storage buffer containing 0.1% sodium azide. Store at -20°C.

ChIP Primers, HoxA2 upstream. One vial containing 75 μL of 5 μM of each primer specific for the promoter region of human HoxA2. Store at -20°C.
FOR: AGG AAA GAT TTT GGT TGG GAA G
REV: AAA AAG AGG GAA AGG GAC AGA C
Format: Purified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Includes negative control mouse IgG antibody and primers specific for human HoxA2 upstream region.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrice Penfornis et al.
Scientific reports, 8(1), 1720-1720 (2018-01-31)
Human mesenchymal stem/stromal cells (hMSCs) provide support for cancer progression, partly through their secretome that includes extracellular vesicles (EVs). Based on deep-sequencing of small RNA from EVs of MSCs, we now report the characterization of novel small RNA, named n-miR-G665
Ritu Kushwaha et al.
Stem cell reports, 6(5), 772-783 (2016-05-03)
Human male germ cell tumors (GCTs) are derived from primordial germ cells (PGCs). The master pluripotency regulator and neuroectodermal lineage effector transcription factor SOX2 is repressed in PGCs and the seminoma (SEM) subset of GCTs. The mechanism of SOX2 repression
Kien Nguyen et al.
mBio, 8(1) (2017-03-02)
We showed previously that the histone lysine methyltransferase (HKMT) H3K27me3 (EZH2) is the catalytic subunit of Polycomb repressive complex 2 (PRC2) and is required for the maintenance of HIV-1 latency in Jurkat T cells. Here we show, by using chromatin
Guo-You Liu et al.
Nucleic acids research, 44(6), 2613-2627 (2015-11-29)
The Hox genes encode transcription factors that determine embryonic pattern formation. In embryonic stem cells, the Hox genes are silenced by PRC2. Recent studies have reported a role for long noncoding RNAs in PRC2 recruitment in vertebrates. However, little is
Malay Mandal et al.
Nature immunology, 12(12), 1212-1220 (2011-11-01)
During B lymphopoiesis, recombination of the locus encoding the immunoglobulin κ-chain complex (Igk) requires expression of the precursor to the B cell antigen receptor (pre-BCR) and escape from signaling via the interleukin 7 receptor (IL-7R). By activating the transcription factor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service