Skip to Content
Merck
All Photos(1)

Key Documents

17-299

Sigma-Aldrich

Anti-Cdc42 Immunoblotting Kit

This Anti-Cdc42 Immunoblotting Kit detects Cdc42 when performing western blots.

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$909.00

$909.00


Estimated to ship on26 May 2025



Select a Size

Change View
1 KIT
$909.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

$909.00


Estimated to ship on26 May 2025


biological source

mouse

Quality Level

antibody product type

primary antibodies

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

Gene Information

human ... CDC42(998)

General description

Anti-cdc42 is a mouse monoclonal IgG1 that recognizes cdc42 from human, mouse, rat and dog

Application

Research Category
Signaling
Research Sub Category
MAP Kinases
This Anti-Cdc42 Immunoblotting Kit detects Cdc42 when performing western blots.

Packaging

Kit capacity: 10 immunoblots per kit

Components

Anti-cdc42, (mouse monoclonal IgG1), Catalog # 05-542

Immunoblot Blocking Reagent (Cat.# 20-200)

Goat Anti-Mouse IgG, HRP conjugate (Cat.# 12-349)

Quality

Routinely evaluated by immunoblot on RIPA lysates from HL-60 cells.

Physical form

See Certificate of Analysis for details

Storage and Stability

Stable for 1 year at -20°C from date of shipment.
Note: Catalog # 12-349, Goat Anti-Mouse IgG, HRP-conjugate is light sensitive, handle accordingly.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela Gismondi et al.
Blood, 104(2), 436-443 (2004-03-06)
In this study we show that Wiskott-Aldrich syndrome protein (WASp), a critical regulator of actin cytoskeleton that belongs to the Scar/WAVE family, plays a crucial role in the control of natural killer (NK) cell cytotoxicity. Analysis of NK cell numbers
Helena Stabile et al.
Blood, 115(14), 2818-2826 (2010-02-05)
We analyzed the involvement of Wiskott-Aldrich syndrome protein (WASp), a critical regulator of actin cytoskeleton remodeling, in the control of natural killer (NK)-cell migration. NK cells derived from patients with Wiskott-Aldrich syndrome/X-linked thrombocytopenia (WAS/XLT), carrying different mutations in the WASP
Thomas Jank et al.
Nature communications, 6, 7807-7807 (2015-07-21)
Yersinia species cause zoonotic infections, including enterocolitis and plague. Here we studied Yersinia ruckeri antifeeding prophage 18 (Afp18), the toxin component of the phage tail-derived protein translocation system Afp, which causes enteric redmouth disease in salmonid fish species. Here we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service