Skip to Content
Merck
All Photos(2)

Documents

172103

Sigma-Aldrich

Anaphase-Promoting Complex Inhibitor Negative Control, AAME

The Anaphase-Promoting Complex Inhibitor Negative Control, AAME, also referenced under CAS 1784-05-0, controls the biological activity of Anaphase-Promoting Complex.

Synonym(s):

Anaphase-Promoting Complex Inhibitor Negative Control, AAME, AAME hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N4O3 · xHCl
CAS Number:
Molecular Weight:
230.26 (free base basis)
UNSPSC Code:
41106609
NACRES:
NA.77

Quality Level

Assay

≥99% (TLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N=C(NCCC[C@@H](C(OC)=O)NC(C)=O)N.[xHCl]

InChI

1S/C9H18N4O3.ClH/c1-6(14)13-7(8(15)16-2)4-3-5-12-9(10)11;/h7H,3-5H2,1-2H3,(H,13,14)(H4,10,11,12);1H/t7-;/m0./s1

InChI key

FIUSXBMWQFBXLZ-FJXQXJEOSA-N

General description

An acetyl-L-arginine analog of TAME that serves as a negative control. TAME is an ubiquitin ligase inhibitor that specifically blocks Anaphase-Promoting Complex (APC) activation.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zeng, X., et al. 2010. Cancer Cell18, 382.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service