Skip to Content
Merck
All Photos(1)

Key Documents

171264

Sigma-Aldrich

AMPK Inhibitor, Compound C•2HCl

InSolution, ≥98%

Synonym(s):

InSolution AMPK Inhibitor, Compound C•2HCl, 6-(4-(2-Piperidin-1-yl-ethoxy)-phenyl)-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine, 2HCl, H₂O, Compound C, 2HCl, H₂O, AMPK Inhibitor I, AMPK Inhibitor I, 6-(4-(2-Piperidin-1-yl-ethoxy)-phenyl)-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine, 2HCl, H₂O, Compound C, 2HCl, H₂O

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$303.00

$303.00


Estimated to ship on16 May 2025



Select a Size

Change View
2 MG
$303.00

About This Item

Empirical Formula (Hill Notation):
C24H25N5O
CAS Number:
Molecular Weight:
399.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$303.00


Estimated to ship on16 May 2025


Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

ambient

storage temp.

−20°C

SMILES string

[n]21ncc(c2ncc(c1)c4ccc(cc4)OCCN5CCCCC5)c3ccncc3

InChI

1S/C24H25N5O/c1-2-12-28(13-3-1)14-15-30-22-6-4-19(5-7-22)21-16-26-24-23(17-27-29(24)18-21)20-8-10-25-11-9-20/h4-11,16-18H,1-3,12-15H2

InChI key

XHBVYDAKJHETMP-UHFFFAOYSA-N

General description

A cell-permeable pyrrazolopyrimidine compound that acts as a potent, selective, reversible, and ATP-competitive inhibitor of AMPK (AMP-activated protein kinase; Ki = 109 nM in the presence of 5 µM ATP and the absence of AMP). Does not affect the activities of ZAPK, Syk, PKCθ, PKA, or JAK3. Blocks celluar activities induced by AICAr (Cat. No.123040) or Metformin (Cat. No. 317240). Induces weight loss by attenuating AMPK-mediated food intake in mice. The solid form of this compound (Cat. No. 171260) is also available.

Packaging

Packaged under inert gas

Physical form

A 10 mM (2 mg/408 µl) solution of AMPK Inhibitor, Compound C, 2HCl, (Cat. No. 171260) in H₂O.

Reconstitution

Following intial thaw, aliquot and freeze (-20°C).

Other Notes

Yu, P.B., et al. 2008. Nat. Chem. Biol4, 33.
Kim, E.K., et al. 2004. J. Biol. Chem.279, 19970.
Lee, M., et al. 2003. J. Biol. Chem.278, 39653.
Inoki, K., et al. 2003. Cell115, 577.
Fryer, L.G., 2002. FEBS Lett.531, 189.
Zhou, G., et al. 2001. J. Clin. Invest.108, 1167.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service