Skip to Content
Merck
All Photos(2)

Key Documents

171262

Sigma-Aldrich

AMPK Activator V, Ampkinone

The AMPK Activator V, Ampkinone modulates the biological activity of AMPK. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

AMPK Activator V, Ampkinone, AKN, AMPK Signaling Activator V

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$275.00

$275.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$275.00

About This Item

Empirical Formula (Hill Notation):
C31H23NO6
CAS Number:
Molecular Weight:
505.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

$275.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 25 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

OC1=C(OC)C=CC2=C1OC(C)(C)C3=C2C=CC4=C3C(N(C5=CC=C(C(C6=CC=CC=C6)=O)C=C5)C4=O)=O

InChI key

BRRCHDIWBKOMEC-UHFFFAOYSA-N

General description

A cell-permeable benzopyran compound that stimulates both LKB1 and CaMKK-mediated AMPK activity indirectly with no noticeable effect on purified AMPK, cellular AMP/ATP ratio, PI 3-K/Akt signaling and respiratory chain, and displays antidiabetic and antiobesity properties. Shown to induce phosphorylation of AMPK-Thr172 and ACC-Ser79 in L6 cells (EC50 = 4.3 µM), C2C12 myocytes, 3T3-L1 preadipocytes and HepG2 hepatic cells, and enhance glucose uptake and insulin sensitivity in high fat-diet-induced obesity mouse model (10 mg/kg, s.c.) with the lowering of cholesterol and nonesterified fatty acids.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Oh, S., et al. 2010. J. Med. Chem.53, 7405.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service