Skip to Content
Merck
All Photos(1)

Key Documents

171255-M

Millipore

Ampicillin, Sodium Salt, Sterile, Tissue Culture Grade

sterile, suitable for tissue culture

Synonym(s):

Ampicillin, Sodium Salt, Sterile, Tissue Culture Grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N3O4S · Na
CAS Number:
Molecular Weight:
371.39
MDL number:
UNSPSC Code:
12352200

sterility

sterile

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

technique(s)

tissue culture: suitable

color

yellow

storage temp.

2-8°C

InChI

1S/C16H19N3O4S.Na/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8;/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23);/q;+1/p-1/t9-,10-,11+,14-;/m1./s1

InChI key

KLOHDWPABZXLGI-YWUHCJSESA-M

General description

A β-lactam antibiotic that inhibits bacterial cell-wall synthesis. Sterilized by γ-irradiation. Potency: ≥ 845 µg/mg.
Antibiotic that kills bacteria by interfering with the terminal reaction in bacterial wall synthesis. Effective against gram-negative bacteria. Ampicillin is ineffective against Pseudomonas, Enterobacter, and indole-positive Proteus infections. Sterilized by γ-irradiation.

Biochem/physiol Actions

Potency (anhydrous): ≥845 µg/mg

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Reconstitute in 20 ml sterile H₂O to yield a final stock concentration of 10 mg/ml.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service