Skip to Content
Merck
All Photos(1)

Key Documents

16-452

Sigma-Aldrich

4G10® Platinum, Anti-Phosphotyrosine Antibody, Biotin Conjugate

clone 4G10®, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,110.00

$1,110.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$1,110.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$1,110.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

4G10®, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG2bκ

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PID1(55022)

General description

The development of the anti-phosphotyrosine, clone 4G10 in 1989 was a monumental discovery for researchers. 4G10 was the first and best single monoclonal antibody for the detection tyrosine phosphorylation. 4G10 is well known for its sensitivity and its ability to detect multiple tyrosine phosphorylations on numerous substrates. It has been validated by thousands of scientific and medical researchers in virtually every application and tyrosine target over the past 2 decades. Now Millipore has made the best even better with 4G10 Platinum. We pooled 4G10 with the next most highly regarded anti-phosphotyrosine, clone PY20, to make 4G10 Platinum. PY20 itself is a very poor substitute for 4G10, but its additive effect allow for a greater level of detection on more substrates that even 4G10 alone is capable.

Immunogen

The immunogen for 4G10 was phospho-tyramine coupled to KLH, while the immunogen for PY20 was phosphotyrosine conjugated to a carrier protein.

Application

4G10 Platinum, Anti-Phosphotyrosine, Biotin Conjugate detects levels of Phosphotyrosine proteins & has been published & validated for use in IP.

Quality

Western Blot Analysis:
A dilution of 1:1000-2000 of this antibody detected tyrosine-phosphorylated proteins in a modified RIPA lysate from human HEK293 cells treated with 100 nM insulin.

Target description

Varies

Physical form

100 μL of a proprietary mixture of biotin conjugated, protein G purified mouse monoclonals 4G10 (IgG2bκ) and PY20 (IgG2b) in PBS, pH 7.4 with 0.05% sodium azide.

Storage and Stability

Stable for 6 months at 2-8°C from date of shipment. For maximum recovery of product, centrifuge the vial prior to removing the cap.
NOTE: DO NOT FREEZE.

Legal Information

4G10 is a registered trademark of Upstate Group, Inc.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Avin Hawez et al.
Laboratory investigation; a journal of technical methods and pathology (2021-11-05)
Sepsis is associated with exaggerated neutrophil responses although mechanisms remain elusive. The aim of this study was to investigate the role of c-Abelson (c-Abl) kinase in neutrophil extracellular trap (NET) formation and inflammation in septic lung injury. Abdominal sepsis was
Ren Liu et al.
Blood, 116(2), 297-305 (2010-05-06)
Axl is an oncogenic receptor tyrosine kinase that plays multiple roles in tumorigenesis and metastasis of many cancers. This study is the first to demonstrate that Axl is induced in Kaposi sarcoma and Kaposi sarcoma herpesvirus (KSHV) transformed endothelial cells.
Kasper J Mygind et al.
International journal of molecular sciences, 25(11) (2024-06-19)
Desmoplasia is a common feature of aggressive cancers, driven by a complex interplay of protein production and degradation. Basigin is a type 1 integral membrane receptor secreted in exosomes or released by ectodomain shedding from the cell surface. Given that
Philippe Foubert et al.
The Journal of clinical investigation, 117(6), 1527-1537 (2007-05-19)
Endothelial progenitor cell (EPC) transplantation has beneficial effects for therapeutic neovascularization; however, only a small proportion of injected cells home to the lesion and incorporate into the neocapillaries. Consequently, this type of cell therapy requires substantial improvement to be of
Shalini Chaturvedi et al.
International journal of biological sciences, 9(10), 1099-1107 (2013-12-18)
A novel assay was developed to measure ratio of p-FMS (phospho FMS) to FMS using the Meso Scale Discovery(®) (MSD) technology and compared to the routinely used, IP-Western based approach. The existing IP-Western assay used lysed PBMCs (Peripheral Blood Mononuclear

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service