Skip to Content
Merck
All Photos(1)

Key Documents

16-254

Sigma-Aldrich

Anti-Histidine Tagged Antibody, clone 4D11, Alexa Fluor 488 conjugate

clone 4D11, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$892.00

$892.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$892.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$892.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4D11, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HRG(3273)

Application

Detect Histidine Tagged using this Anti-Histidine Tagged Antibody (anti His antibody), clone 4D11, Alexa Fluor 488 conjugate validated for use in IF, WB & IC.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Quality

Routinely evaluated by immunoblot on recombinant His protein

Storage and Stability

stable 1 year at 4°C from date of shipment

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C S McPhee et al.
Journal of dairy science, 94(4), 1738-1743 (2011-03-24)
Despite the recent growth of the organic dairy industry, organic producers and veterinarians have limited information when choosing mastitis treatments for animals in organic dairy production. Organic producers commonly administer homeopathic or other plant-based products without having research evaluating the
Michael Helwig et al.
The Journal of biological chemistry, 288(2), 1114-1124 (2012-11-23)
Neurodegenerative diseases such as Alzheimer (AD) and Parkinson (PD) are characterized by abnormal aggregation of misfolded β-sheet-rich proteins, including amyloid-β (Aβ)-derived peptides and tau in AD and α-synuclein in PD. Correct folding and assembly of these proteins are controlled by
Saraswoti Khadge et al.
Journal of mammary gland biology and neoplasia, 23(1-2), 43-58 (2018-03-27)
Studies in rodents have shown that dietary modifications as mammary glands (MG) develop, regulates susceptibility to mammary tumor initiation. However, the effects of dietary PUFA composition on MGs in adult life, remains poorly understood. This study investigated morphological alterations and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service