Skip to Content
Merck
All Photos(1)

Key Documents

16-218

Sigma-Aldrich

Anti-Phospho-Histone H3 (Ser10) Antibody, clone 3H10, Cy5 conjugate

clone 3H11, Upstate®, from mouse

Synonym(s):

H3 histone, family 3A, H3S10P, Histone H3 (phospho S10), H3 histone, family 3A, H3 histone, family 3B, H3 histone, family 3B (H3.3B)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$1,030.00

$1,030.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$1,030.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$1,030.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

CY5 conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

3H11, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer10)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure.

The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

Expected to cross react with all vertebrate species.
Histone H3 phosphorylated at Ser10

Immunogen

A proprietary immunogen based on a peptide sequence containing phospho-serine corresponding to residue 10 of human histone H3, clone 3H10. This is a Cy5 conjugate of Anti-phospho-Histone H3 (Ser10), clone 3H10, Catalog # 05-806.

Application

Anti-Phospho-Histone H3 (Ser10) Antibody, clone 3H10, Cy5 Conjugate is a Mouse Monoclonal Antibody for detection of Histone H3 phosphorylated at serine 10. This purified mAb, also known as H3S10p,
is labeled with Cy5.5 NHS ester (Cy5) and validated in IF, WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Routinely evaluated by immunoblot in acid extrated protein from mitotic HeLa cells treated with colcemid (17-306).

Target description

17kDa

Physical form

Protein G Purified

Storage and Stability

1 year at 4°C from date of shipment

Analysis Note

Control
Acid extracts from colcemid treated HeLa cells (lane 2)

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorge Boucas et al.
PloS one, 10(5), e0125745-e0125745 (2015-05-21)
Growing evidence suggests a key role for RNA binding proteins (RBPs) in genome stability programs. Additionally, recent developments in RNA sequencing technologies, as well as mass-spectrometry techniques, have greatly expanded our knowledge on protein-RNA interactions. We here use full transcriptome

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service