Skip to Content
Merck
All Photos(1)

Key Documents

16-170

Sigma-Aldrich

Anti-Myc Tag Antibody, clone 9E10, biotin conjugate

clone 9E10, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
$880.00

$880.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
$880.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$880.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

9E10, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

General description

Anti-Myc Tag antibody (05-419) conjugated to biotin

Application

Anti-Myc Tag Antibody, clone 9E10, biotin conjugate detects level of Myc Tag & has been published & validated for use in WB & IC.
For use in applications in which a biotin conjugate is advantageous, such as WB and ICC.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Quality

routinely evaluated by immunoblot in HeLa nuclear extract and myc-tagged recombinant protein

Physical form

PBS and 0.05% sodium azide

Storage and Stability

1 year at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manas Chakraborty et al.
Methods in cell biology, 144, 307-327 (2018-05-29)
During mitosis, kinetochores often bind to the walls of spindle microtubules, but these lateral interactions are then converted into a different binding mode in which microtubule plus-ends are embedded at kinetochores, forming dynamic "end-on" attachments. This remarkable configuration allows continuous
Sandra E Echeverría et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 18(11), 3024-3029 (2009-10-22)
Few studies have examined the role of neighborhood socioeconomic condition in shaping breast cancer disparities in defined local areas. We tested associations between three measures of neighborhood socioeconomic condition (poverty, median income, and a composite neighborhood score) on breast cancer
T A Endo et al.
Nature, 387(6636), 921-924 (1997-06-26)
The proliferation and differentiation of cells of many lineages are regulated by secreted proteins known as cytokines. Cytokines exert their biological effect through binding to cell-surface receptors that are associated with one or more members of the JAK family of
Michael W Linhoff et al.
Neuron, 61(5), 734-749 (2009-03-17)
Delineating the molecular basis of synapse development is crucial for understanding brain function. Cocultures of neurons with transfected fibroblasts have demonstrated the synapse-promoting activity of candidate molecules. Here, we performed an unbiased expression screen for synaptogenic proteins in the coculture
O Zugasti et al.
Molecular and cellular biology, 21(19), 6706-6717 (2001-09-05)
Signals from the extracellular matrix are essential for the survival of many cell types. Dominant-negative mutants of two members of Rho family GTPases, Rac1 and Cdc42, mimic the loss of anchorage in primary mouse fibroblasts and are potent inducers of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service