Skip to Content
Merck
All Photos(1)

Key Documents

1.51252

Supelco

Coupling unit (dead volume-free) for capillaries 1/16" or 0.5 mm

Sign Into View Organizational & Contract Pricing

Select a Size

3 UNITS
$617.00

$617.00


Estimated to ship on21 May 2025



Select a Size

Change View
3 UNITS
$617.00

About This Item

UNSPSC Code:
41115716

$617.00


Estimated to ship on21 May 2025


technique(s)

HPLC: suitable

Quality Level

storage temp.

no temp limit

General description

The package contains: 3 pieces.

Analysis Note

visual control: passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service